Skip to Content
Merck
All Photos(1)

Key Documents

36664

Supelco

Aldrin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6
CAS Number:
Molecular Weight:
364.91
Beilstein:
2336652
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)[C@]2(Cl)[C@@H]3[C@@H]4C[C@@H](C=C4)[C@@H]3[C@@]1(Cl)C2(Cl)Cl

InChI

1S/C12H8Cl6/c13-8-9(14)11(16)7-5-2-1-4(3-5)6(7)10(8,15)12(11,17)18/h1-2,4-7H,3H2/t4-,5+,6+,7-,10+,11-

InChI key

QBYJBZPUGVGKQQ-SJJAEHHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aldrin belongs to the class of organochlorine pesticides, commonly used in agriculture and is persistent as a highly toxic environmental contaminant.

Application

Aldrin may be used as an analytical reference standard for the quantification of the analyte in environmental samples and animal fats using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-tube solid-phase microextraction-capillary liquid chromatography as a solution for the screening analysis of organophosphorus pesticides in untreated environmental water samples
Chafer-Pericas.C, et al.
Journal of Chromatography A, 1141(1), 10-21 (2007)
A toxic reagent-free method for normal-phase matrix solid-phase dispersion extraction and reversed-phase liquid chromatographic determination of aldrin, dieldrin, and DDTs in animal fats
Furusawa.N, et al.
Analytical and Bioanalytical Chemistry, 378(8) (2004)
Determination of organochlorine and pyrethroid pesticides in fruit and vegetables using solid phase extraction clean-up cartridges.
Sharif Z, et al.
Journal of Chromatography A, 1127(1-2), 254-261 (2006)
Song-Yee Baek et al.
Environmental science & technology, 45(10), 4475-4482 (2011-04-15)
XAD-2 resin based passive air samplers (PAS) were deployed for three one-year periods at the Korean polar and South Pacific research stations at Ny-Ålesund (2005-2009), King George Island (2005-2007), and Chuuk (2006-2009) to investigate long-range transport, local sources, and temporal
Xiaomin Li et al.
Journal of chromatography. A, 1277, 69-75 (2013-01-22)
Identification and quantification of related-structure impurity is a research focus in the purity assessment of organic compounds. Determination of the purity value and uncertainty assessment are also important in the metrological research. A method for the determination of related-structure impurity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service