Skip to Content
Merck
All Photos(1)

Documents

35800

Supelco

Biphenyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21
Beilstein:
1634058
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.31 (vs air)

vapor pressure

9.46 mmHg ( 115 °C)

product line

PESTANAL®

autoignition temp.

1004 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.6 %, 111 °F
5.8 %, 166 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

255 °C (lit.)

mp

68-70 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

c1ccc(cc1)-c2ccccc2

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Biphenyl may also serve as a polymerizable electrolyte additive for overcharge protection of lithium ion batteries.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical behavior of biphenyl as polymerizable additive for overcharge protection of lithium ion batteries
Xiao L, et al.
Electrochimica Acta, 49, 4189-4196 (2004)
Xiaofeng Chen et al.
The Journal of organic chemistry, 77(17), 7572-7578 (2012-08-07)
A highly efficient cross-coupling of diarylborinic acids and anhydrides with aryl chlorides and bromides has been effected by using a palladium catalyst system co-supported by a strong σ-donor N-heterocyclic carbene (NHC), N,N'-bis(2,6-diisopropylphenyl) imidazol-2-ylidene, and a strong π-acceptor phosphite, triphenylphosphite, in
Yang Ou et al.
Chemical communications (Cambridge, England), 49(33), 3473-3475 (2013-03-22)
A novel A21-CuI catalyzed direct nitrogenation of biphenyl halides for the direct synthesis of carbazoles via a direct C-H amination process has been developed. A recyclable and inexpensive Cu-catalyst was successfully employed in N-heterocyclic compound synthesis via tandem azidation and
Guoxiang Jiang et al.
Carbohydrate polymers, 92(1), 758-764 (2012-12-12)
Ultrasound-assisted extraction was employed to extract polysaccharides from longan seed (LSP), with aids of a Box-Behnken statistical design to investigate the effects of ultrasonic power, time and liquid/solid ratio on the extraction recovery of the LSP. The structural analysis indicated
Frédéric R Leroux et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(45), 14232-14236 (2012-09-29)
A modular way towards biaryls: Highly enantioenriched biphenyls can be prepared based on a transition-metal-free aryl-aryl coupling followed by efficient desymmetrization or deracemization and chemoselective functionalization (see scheme).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service