Skip to Content
Merck
All Photos(2)

Key Documents

35394

Supelco

Albendazole sulfone

VETRANAL®, analytical standard

Synonym(s):

Methyl [5-(propylsulfonyl)-1H-benzimidazol-2-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O4S
CAS Number:
Molecular Weight:
297.33
Beilstein:
8394880
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCS(=O)(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O4S/c1-3-6-20(17,18)8-4-5-9-10(7-8)14-11(13-9)15-12(16)19-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

CLSJYOLYMZNKJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Albendazole sulfone is a nonactive metabolite of albendazole sulfoxide (ABZSO).

Application

Albendazole sulfone may be used as a reference standard in the determination of albendazole sulfone in plasma samples using high-performance liquid chromatography (HPLC) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,spleen,male reproductive organs,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Constant Edi et al.
PLoS neglected tropical diseases, 13(5), e0007325-e0007325 (2019-05-21)
A single co-administered dose of ivermectin (IVM) plus diethylcarbamazine (DEC) plus albendazole (ALB), or triple-drug therapy, was recently found to be more effective for clearing microfilariae (Mf) than standard DEC plus ALB currently used for mass drug administration programs for
Victoria Miró et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(4), 408-414 (2019-07-16)
Combinations of bioactive phytochemicals with synthetic compounds have been suggested as promissory tools for the improvement of nematode control in livestock. Bioactive phytochemicals may interfere with the activity of drug-metabolizing enzymes and delay the metabolic conversion of anthelmintics into less
Jessica D Schulz et al.
Antimicrobial agents and chemotherapy, 63(4) (2019-02-13)
Albendazole is an effective anthelmintic intensively used for decades. However, profound pharmacokinetic (PK) characterization is missing in children, the population mostly affected by helminth infections. Blood microsampling would facilitate PK studies in pediatric populations but has not been applied to
Huan Sun et al.
Journal of agricultural and food chemistry, 64(1), 356-363 (2015-12-15)
A novel nickel oxide nanoparticle-deposited silica (SiO2@NiO) composite was prepared via liquid-phase deposition (LPD) and then employed as a solid-phase extraction (SPE) sorbent. When the SPE was coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI/MS) analysis, an analytical platform for
Francesca Accioni et al.
Journal of agricultural and food chemistry, 67(1), 520-530 (2018-12-06)
In this work, a restricted access volatile supramolecular solvent (RAM-VOL-SUPRAS) directly synthesized in milk is proposed for the first time for the simultaneous extraction and cleanup of benzimidazole anthelmintic drugs in milk meant for human consumption. The RAM-VOL-SUPRAS was formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service