Skip to Content
Merck
All Photos(2)

Key Documents

35391

Supelco

Triclabendazole sulfoxide

VETRANAL®, analytical standard

Synonym(s):

5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl3N2O2S
CAS Number:
Molecular Weight:
375.66
Beilstein:
5829656
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CS(=O)c1nc2cc(Cl)c(Oc3cccc(Cl)c3Cl)cc2[nH]1

InChI

1S/C14H9Cl3N2O2S/c1-22(20)14-18-9-5-8(16)12(6-10(9)19-14)21-11-4-2-3-7(15)13(11)17/h2-6H,1H3,(H,18,19)

InChI key

GABQPFWIQFRJSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triclabendazole sulfoxide belongs to the benzimidazole class of anthelmintics drugs. It is very much effective against mature and immature stages of Fasciola hepatica and Fasciola gigantica in all ruminant species.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Keiser et al.
The American journal of tropical medicine and hygiene, 75(2), 287-291 (2006-08-10)
Some have claimed that triclabendazole, a safe and efficacious drug for the treatment of fascioliasis, also exhibits antischistosomal properties, but results are conflicting. We assessed the effect of triclabendazole and its two main metabolites against two different strains of Schistosoma
L Mottier et al.
Experimental parasitology, 113(1), 49-57 (2006-01-25)
Anthelmintic molecules must reach their receptors inside target parasites to exert the pharmacological effect. Available data suggest that the main route of entry of antiparasitic drugs into helminth parasites would be through their external surface. However, it is unclear if
I Fairweather et al.
Veterinary parasitology, 183(3-4), 249-259 (2011-08-09)
The aim of this study was to develop an Egg Hatch Assay (EHA) test for the detection of triclabendazole (TCBZ) resistance in Fasciola hepatica. A number of fluke isolates were used, of differing sensitivity to TCBZ. Eggs were exposed to
A W Stitt et al.
Parasitology research, 82(4), 333-339 (1996-01-01)
The effects of the active sulphoxide metabolite of the fasciolicide triclabendazole (Fasinex, Ciba-Geigy) on the vitelline cells of Fasciola hepatica were determined in vitro by transmission electron microscopy using both intact flukes and tissue-slice material. At a triclabendazole concentration of
Catherine Devine et al.
Parasitology research, 106(6), 1351-1363 (2010-03-26)
A study has been carried out to investigate whether the action of triclabendazole (TCBZ) against Fasciola hepatica is altered by inhibition of drug metabolism. The flavin monooxygenase system (FMO) was inhibited using methimazole (MTZ) to see whether a TCBZ-resistant isolate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service