Skip to Content
Merck
All Photos(2)

Key Documents

34221

Supelco

2-Isopropylthioxanthone

PESTANAL®, analytical standard

Synonym(s):

2-Isopropyl-9H-thioxanthen-9-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14OS
CAS Number:
Molecular Weight:
254.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

CC(C)c1ccc2Sc3ccccc3C(=O)c2c1

InChI

1S/C16H14OS/c1-10(2)11-7-8-15-13(9-11)16(17)12-5-3-4-6-14(12)18-15/h3-10H,1-2H3

InChI key

KTALPKYXQZGAEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Isopropylthioxanthone is a compound, which can play the role of a sulfur type photoinitiator in printing industries and UV-cured inks for food packaging materials.

Application

2-Isopropylthioxanthone has been used as an analytical standard for the determination of the analyte in packaged food materials using liquid chromatography-tandem mass spectrometry (LC–MS/MS). It may be used as an analytical standard for the determination of the analyte in milk, fruit drinks and packaged beverages by high performance liquid chromatography (HPLC) coupled to MS/MS and micellar electrokinetic chromatography techniques (MEKC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaotian Zhao et al.
Journal of molecular modeling, 26(3), 56-56 (2020-02-13)
The activation or functionalization of the saturated C-H is an extremely active field at present. We have explored the triplet state thioxanthone in reactivity of the hydrogen transfer reaction between donors and acceptors. In our works, two donors with quasi-inert
Ye Fan et al.
Colloids and surfaces. B, Biointerfaces, 167, 385-391 (2018-04-28)
FAVs (fatty acid vesicles), originated from natural biomass and created available biointerface, have advantages of biocompatibility, low-cost, and easy self-assembly in aqueous solution due to their dynamic feature. However, there is no example of applying FAVs in contact with human
Direct analysis of isopropylthioxanthone (ITX) in milk by high?performance liquid chromatography/tandem mass spectrometry
Bagnati R, et al.
Rapid Communications in Mass Spectrometry, 21, 1998-2002 (2007)
AhR-agonistic, anti-androgenic, and anti-estrogenic potencies of 2-isopropylthioxanthone (ITX) as determined by in vitro bioassays and gene expression profiling
Peijnenburg.A, et al.
Toxicology in vitro, 24, 1619-1628 (2010)
Gianni Sagratini et al.
Journal of chromatography. A, 1194(2), 213-220 (2008-05-20)
A new analytical method, using gas chromatography-mass spectrometry (GC/MS) and liquid chromatography-mass spectrometry (LC/MS) techniques, was developed for the determination in packaged food beverages of five ink photoinitiator residues: 2-isopropylthioxanthone (ITX), benzophenone, 2-ethylhexyl-4-dimethylaminobenzoate (EHDAB), 1-hydroxycyclohexyl-1-phenyl ketone (IRGACURE 184) and ethyl-4-dimethylaminobenzoate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service