Skip to Content
Merck
All Photos(2)

Key Documents

33970

Supelco

Mepanipyrim

PESTANAL®, analytical standard

Synonym(s):

4-Methyl-N-phenyl-6-(1-propynyl)-2-pyrimidinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3
CAS Number:
Molecular Weight:
223.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC#Cc1cc(C)nc(Nc2ccccc2)n1

InChI

1S/C14H13N3/c1-3-7-13-10-11(2)15-14(17-13)16-12-8-5-4-6-9-12/h4-6,8-10H,1-2H3,(H,15,16,17)

InChI key

CIFWZNRJIBNXRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

267.8 °F

Flash Point(C)

131 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Virgínia C Fernandes et al.
Environmental science and pollution research international, 19(9), 4184-4192 (2012-05-09)
Pesticides are among the most widely used chemicals in the world. Because of the widespread use of agricultural chemicals in food production, people are exposed to low levels of pesticide residues through their diets. Scientists do not yet have a
Michiko Nakamura et al.
Bioscience, biotechnology, and biochemistry, 67(1), 139-150 (2003-03-07)
Mepanipyrim inhibited retrograde Golgi-to-ER trafficking induced by brefeldin A (BFA), nordihydroguaiaretic acid, clofibrate, and arachidonyltrifluoromethyl ketone in NRK and other types of cells, but did not inhibit anterograde trafficking of Golgi-resident proteins translocated to ER by BFA and newly synthesized
Paola Calza et al.
Journal of chromatography. A, 1049(1-2), 115-125 (2004-10-27)
Mepanipyrim, a widely used pyrimidinic fungicide, has been photocatalytically degraded in aqueous solution on TiO2. The purpose of this study is to artificially produce degradation compounds similar to those formed in the environment. Numerous intermediates have been identified and characterised
M Terada et al.
The Journal of toxicological sciences, 23(3), 223-234 (1998-10-21)
Mepanipyrim, a new fungicide, was administered orally to rats, mice and dogs for 13 weeks to clarify its toxic profiles. Hepatotoxicities were observed characteristically in these species with high concentrations of mepanipyrim; more than 200 ppm in rats, more than
Toshihiro Nagata et al.
Pest management science, 60(4), 399-407 (2004-05-04)
A series of 2-anilinopyrimidines was prepared and their fungicidal activities against Botrytis cinerea Pers were examined. The activity fell sharply with any substitution on the anilinobenzene ring. Substituents at the 5-position of the pyrimidine ring greatly reduced the activity. Substituents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service