Skip to Content
Merck
All Photos(1)

Key Documents

33824

Supelco

Pentadecafluorooctanoic acid

analytical standard

Synonym(s):

PFAS, PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)6COOH
CAS Number:
Molecular Weight:
414.07
Beilstein:
1809678
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101

reg. compliance

suitable for FDA C-010.02

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

189 °C/736 mmHg (lit.)

mp

55-56 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

application(s)

PFAS testing
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

InChI

1S/C8HF15O2/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23/h(H,24,25)

InChI key

SNGREZUHAYWORS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentadecafluorooctanoic acid is a perfluoroalkyl carboxylic acid, which is primarily used as an ion pair reagent.

Application

Pentadecafluorooctanoic acid may be used as an ion pair reagent in the determination of underivatized amino acids in fermentation media using liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - Skin Corr. 1C - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of underivatized pyroglutamic acid, glutamic acid, glutamine and other relevant amino acids in fermentation media by LC-MS-MS.
Qu J, et al.
Analyst, 127(1), 66-69 (2002)
Ion-pair reversed-phase liquid chromatography for determination of polar underivatized amino acids using perfluorinated carboxylic acids as ion pairing agent.
Petritis NK, et al.
Journal of Chromatography A, 833(2), 147-155 (1999)
Anne Vested et al.
Environmental health perspectives, 121(4), 453-458 (2013-01-31)
Perfluorinated alkyl acids (PFAAs), persistent chemicals with unique water-, dirt-, and oil-repellent properties, are suspected of having endocrine-disrupting activity. The PFAA compounds perfluorooctanoic acid (PFOA) and perfluorooctane sulfonic acid (PFOS) are found globally in humans; because they readily cross the
A K Rosenmai et al.
Toxicology and applied pharmacology, 266(1), 132-142 (2012-11-13)
Polyfluoroalkyl phosphate surfactants (PAPS) are widely used in food contact materials (FCMs) of paper and board and have recently been detected in 57% of investigated materials. Human exposure occurs as PAPS have been measured in blood; however knowledge is lacking
Mei-Huei Chen et al.
Epidemiology (Cambridge, Mass.), 24(6), 800-808 (2013-09-17)
Epidemiologic data regarding the potential neurotoxicity of perfluorinated compounds (PFCs) are inconclusive. We investigated the associations between in utero exposure to perfluorooctanoic acid (PFOA) and perfluorooctyl sulfonate (PFOS) and early childhood neurodevelopment. We recruited 239 mother-infant pairs in northern Taiwan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service