33565
4-tert-Octylphenol-ring-13C6 solution
10 μg/mL in acetone, analytical standard
Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol-13C6, 4-tert-OP-13C6
About This Item
Recommended Products
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
concentration
10 μg/mL in acetone
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
mass shift
M+6
storage temp.
−20°C
SMILES string
O[13C]1=[13CH][13CH]=[13C](C(C)(C)CC(C)(C)C)[13CH]=[13CH]1
InChI
1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3/i6+1,7+1,8+1,9+1,11+1,12+1
InChI key
ISAVYTVYFVQUDY-UJYFPCGESA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
1.4 °F - Information refers to the main ingredient.
Flash Point(C)
-17 °C - Information refers to the main ingredient.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service