Skip to Content
Merck
All Photos(1)

Documents

33380

Sigma-Aldrich

4-Methyl-o-phenylenediamine

purum, ≥98.0% (NT)

Synonym(s):

3,4-Toluenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
Beilstein:
507965
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (NT)

bp

155-156 °C/18 mmHg (lit.)

mp

87-89 °C (lit.)
87-89 °C

application(s)

agriculture
environmental

SMILES string

Cc1ccc(N)c(N)c1

InChI

1S/C7H10N2/c1-5-2-3-6(8)7(9)4-5/h2-4H,8-9H2,1H3

InChI key

DGRGLKZMKWPMOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-o-phenylenediamine is the 4-substituted derivative of o-phenylenediamine. It reacts with selenous acid in acid solution to form benzoselenadiazoles.

Application

4-Methyl-o-phenylenediamine was employed as chromogenic reagent in the spectrophotometric determination of selenium(IV). It was used in the synthesis of an asymmetrical tetradentate Schiff base via condensation with dehydroacetic acid and salicylic aldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and thermal study of some transition metal complexes of an asymmetrical tetradentate Schiff base ligand.
Munde AS, et al.
J. Serb. Chem. Soc., 75(3), 349-359 (2010)
Spectrophotometric determination of selenium (IV) with 4-methyl-o-phenylenediamine based on piazselenol formation.
Kartal S, et al.
Journal of Analytical Chemistry, 65(12), 1221-1227 (2010)
Some 4-substituted< i> o</i>-phenylenediamines as reagents for selenium.
Tanaka M and Kawashima T.
Talanta, 12(3), 211-219 (1965)
T A Marks et al.
Teratology, 24(3), 253-265 (1981-12-01)
Pregnant outbred albino (CD-1) mice were given 2-nitro-p-phenylenediamine (2NPPD; 32-256 mg/kg/day), 4-nitro-o-phenylenediamine (4NOPD; 16-1024 mg/kg/day) or 2,5-toluenediamine sulfate (2,5TDS; 16-64 mg/kg/day) by subcutaneous injection on Days 6-15 of gestation. The mice were killed on Day 18, the general health and
Effect of 4 toluene diamine isomers on murine testicular DNA synthesis.
E J Greene et al.
Mutation research, 91(1), 75-79 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service