Skip to Content
Merck
All Photos(2)

Documents

32999

Supelco

Aminoethoxyvinyl glycine hydrochloride

PESTANAL®, analytical standard

Synonym(s):

(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, L-α-(2-Aminoethoxyvinyl)glycine hydrochloride, AVG-HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O3 · HCl
CAS Number:
Molecular Weight:
196.63
Beilstein:
6715149
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

Cl[H].NCCO\C=C\[C@H](N)C(O)=O

InChI

1S/C6H12N2O3.ClH/c7-2-4-11-3-1-5(8)6(9)10;/h1,3,5H,2,4,7-8H2,(H,9,10);1H/b3-1+;/t5-;/m0./s1

InChI key

ZDCPLYVEFATMJF-BTIOQYSDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Inhibition of ethylene biosynthesis delays ripening of fruits in plants treated with AVG.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Yoram Kapulnik et al.
Journal of experimental botany, 62(8), 2915-2924 (2011-02-11)
Strigolactones (SLs) or derivatives thereof have been identified as phytohormones, and shown to act as long-distance shoot-branching inhibitors. In Arabidopsis roots, SLs have been suggested to have a positive effect on root-hair (RH) elongation, mediated via the MAX2 F-box. Two
Chenshuo Chang et al.
Journal of plant physiology, 167(14), 1152-1156 (2010-07-08)
Ethylene is an important plant gas hormone, and the amino acid Glu is emerging as a messenger molecule in plants. To evaluate the role of ethylene and Glu in seed germination and radicle growth under salt stress, effects of 1-aminocyclopropane-1-carboxylic
S Ali et al.
Journal of applied microbiology, 113(5), 1139-1144 (2012-07-24)
The ability of 1-aminocyclopropane-1-carboxylate (ACC) deaminase-containing plant growth-promoting bacterial (PGPB) endophytes Pseudomonas fluorescens YsS6 and Pseudomonas migulae 8R6, their ACC deaminase minus mutants and the rhizospheric plant growth-promoting bacterium Pseudomonas putida UW4 to delay the senescence of mini carnation cut
Naoshi Kojima et al.
Bioorganic & medicinal chemistry letters, 19(8), 2144-2147 (2009-03-24)
Solid-support conjugation at the 5'-terminal primary amine of oligonucleotides is a convenient and powerful method for introducing various functional groups. However, conventional aliphatic amines do not necessarily provide conjugates with sufficient yields. To improve the modification efficacy, we used the
Liangsheng Wang et al.
Plant & cell physiology, 53(6), 1093-1105 (2012-05-25)
When plants are subjected to a deficiency in inorganic phosphate (Pi), they exhibit an array of responses to cope with this nutritional stress. In this work, we have characterized two Arabidopsis mutants, hps3-1 and hps3-2 (hypersensitive to Pi starvation 3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service