Skip to Content
Merck
All Photos(1)

Key Documents

32435

Supelco

Saflufenacil

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-{[methyl(1-methylethyl)amino]sulfonyl}benzamide, N′-{2-Chloro-4-fluoro-5-[1,2,3,6-tetrahydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzoyl}-N-isopropyl-N-methylsulfamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClF4N4O5S
CAS Number:
Molecular Weight:
500.85
Beilstein:
11326826
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)N(C)S(=O)(=O)NC(=O)c1cc(N2C(=O)C=C(N(C)C2=O)C(F)(F)F)c(F)cc1Cl

InChI

1S/C17H17ClF4N4O5S/c1-8(2)25(4)32(30,31)23-15(28)9-5-12(11(19)6-10(9)18)26-14(27)7-13(17(20,21)22)24(3)16(26)29/h5-8H,1-4H3,(H,23,28)

InChI key

GNHDVXLWBQYPJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Saflufenacil is a new pyrimidinedione herbicide registered for the preplant burndown and preemergence use in various agronomic crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Saflufenacil may be used as an analytical reference standard for the determination of the analyte in:
  • Crop plants by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and ultra-high performance liquid chromatography-electrospray ionization-tandem mass spectrometry (UHPLC-ESI-MS/MS) with selected reaction monitoring (SRM) detection.
  • Maize grain and plant samples by modified QuEChERS extraction and LC-MS/MS.
  • Soil samples by modified QuEChERS extraction and UPLC-MS/MS.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Wu et al.
Journal of food biochemistry, 43(4), e12778-e12778 (2019-07-30)
A reliable and efficient method was firstly established for the simultaneous determination of saflufenacil and its metabolites (M800H02, M800H11 and M800H35) in cereals (soybean and corn) and fruits (apple, grape and orange), based on a triple quadrupole liquid chromatography mass
Grain sorghum response to saflufenacil applied preemergence.
Keeling JW, et al.
Crop Protection, 46, 1-6 (2013)
Multiresidue determination of pesticides in crop plants by the quick, easy, cheap, effective, rugged, and safe method and ultra-high-performance liquid chromatography tandem mass spectrometry using a calibration based on a single level standard addition in the sample.
Viera MS, et al.
Journal of Chromatography A, 1526(2), 119-127 (2017)
Simultaneous determination of saflufenacil and its two metabolites in soil samples by ultra-performance liquid chromatography with tandem mass spectrometry.
Lan M, et al.
Journal of Separation Science, 37(6), 658-664 (2014)
Simultaneous Determination of Dimethenamid, Saflufenacil and their Metabolites in Maize Using a Modified QuEChERS Method and Liquid Chromatography-Tandem Mass Spectrometry.
Chen H, et al.
Food Analytical Methods, 11(12), 3396-3405 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service