Skip to Content
Merck
All Photos(1)

Documents

32018

Supelco

1,4-Diethylbenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(C2H5)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903396
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

0.99 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

806 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.495 (lit.)
n20/D 1.495

bp

184 °C (lit.)

mp

−43 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCc1ccc(CC)cc1

InChI

1S/C10H14/c1-3-9-5-7-10(4-2)8-6-9/h5-8H,3-4H2,1-2H3

InChI key

DSNHSQKRULAAEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Diethylbenzene is one of the three isomers of diethylbenzene, which is primarily used as a cross-linker for the fabrication of synthetic rubbers, polyester resins, ion-exchange resins, etc.

Application

1,4-Diethylbenzene may be used as an analytical reference standard for the determination of the analyte in gasoline samples, crude oil and insecticidal formulations by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F

Flash Point(C)

56 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of aromatic hydrocarbons in gasolines by gas chromatography/Fourier transform infrared spectroscopy
Diehl JW, et al.
Analytical Chemistry, 67(13), 2015-2019 (1995)
Olfactory mucosal necrosis in rats following acute intraperitoneal administration of 1, 2-diethylbenzene, 1, 2-diacetylbenzene and 2, 5-hexanedione
Gagnaire F and Boucard S
Neurotoxicology, 41(12), 9-19 (2014)
High pressure liquid chromatographic analysis of chlorpyrifos containing insecticidal formulations: collaborative study
Skelly NE, et al.
Journal of the Association of Official Analytical Chemists, 64(3), 628-634 (1981)
Determination of Hydrocarbons in Crude Oil by Capillary-Column Gas Chromatography.
Martin RL and Winters JC
Analytical Chemistry, 35(12), 1930-1933 (1963)
Analysis of aromatic compounds in gasoline with flow-switching comprehensive two-dimensional gas chromatography.
Micyus NJ, et al.
Journal of Chromatography A, 1086(1-2), 115-121 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service