Skip to Content
Merck
All Photos(3)

Key Documents

319279

Sigma-Aldrich

Phosphomolybdic acid solution

20 wt. % in ethanol

Synonym(s):

Molybdophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(Mo3O10)4]
CAS Number:
Molecular Weight:
1825.25
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

reaction suitability

reagent type: oxidant

concentration

20 wt. % in ethanol

density

0.94 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.OP(O)(O)=O

InChI

1S/12Mo.H3O4P.36O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

DHRLEVQXOMLTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphomolybdic acid solution has been used in the preparation of supported phosphomolybdic acid membranes for use as superacid membranes for the oxidation of lighter alkanes in a catalytic membrane reactor.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Ox. Liq. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

55.0 °F - closed cup

Flash Point(C)

12.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Innovative membrane based process for the selective oxidation of light alkanes assisted by the Fenton
Espro, C, et al.
Asia-Pacific Journal of Chemical Engineering, 5(1), 66-72 (2010)
Maher Mahmoud El-Hashash et al.
Acta pharmaceutica (Zagreb, Croatia), 60(3), 361-367 (2010-12-08)
In the present study, antioxidant activity of methanolic extracts of the leaves of seven Egyptian Cassia species was investigated using two methods, the phosphomolyb-date method and 1,1 diphenyl-2-picrylhydrazyl radical (DPPH) scavenging activity method.· The results revealed that C. glauca is
Rubel Chakravarty et al.
Journal of chromatography. A, 1220, 82-91 (2011-12-27)
Barium-137m availed from a (137)Cs/(137m)Ba generator is an attractive industrial radiotracer for liquid flow rate measurement owing to its short half-life (T(1/2)=2.5 min) and emission of highly energetic gamma radiation (E(γ)=661.7 keV). Ammonium molybdophosphate (AMP) impregnated alumina microspheres (AMP-Al(2)O(3)), an
Dawei Fan et al.
Journal of colloid and interface science, 351(1), 151-155 (2010-08-19)
Hybrid films composed of oppositely charged Keggin-type 12-molybdophosphoric acid H(3)PMo(12)O(40).nH(2)O (PMo(12)) and water-soluble cationic meso-tetra (N-methyl-4-pyridyl) porphinetetratosylate (TMPyP) were fabricated onto silicon and ITO substrates by a layer-by-layer (LbL) self-assembly technique. The LbL films were characterized by UV-vis spectroscopy, X-ray
Juan Feng et al.
Analytical biochemistry, 409(1), 144-149 (2010-10-26)
The classical malachite green (MLG) assay of phosphate, which added MLG after molybdate to the acidified reaction solutions of phosphate, tolerated interference from papaverine, sildenafil, and some similar hydrophobic amines. Resonance Rayleigh scattering signals, the alleviation of interference by poly(vinyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service