Skip to Content
Merck
All Photos(1)

Key Documents

31662

Supelco

Lufenuron

PESTANAL®, analytical standard

Synonym(s):

1-[2,5-Dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]-3-(2,6-difluorobenzoyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H8Cl2F8N2O3
CAS Number:
Molecular Weight:
511.15
Beilstein:
8398291
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

FC(C(F)(F)F)C(F)(F)Oc1cc(Cl)c(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl

InChI

1S/C17H8Cl2F8N2O3/c18-6-5-11(32-17(26,27)14(22)16(23,24)25)7(19)4-10(6)28-15(31)29-13(30)12-8(20)2-1-3-9(12)21/h1-5,14H,(H2,28,29,30,31)

InChI key

PWPJGUXAGUPAHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lufenuron is an insect growth regulator belonging to the benzoylphenylurea class of insecticide. It is used in controlling Ctenocephalides felis (cat flea) in cats.

Application

Lufenuron may be used as a reference standard in the determination of lufenuron in wheat flour using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>338.0 °F - (External MSDS)

Flash Point(C)

> 170 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyp12a4 confers lufenuron resistance in a natural population of Drosophila melanogaster
Bogwitz R M, et al.
Proceedings of the National Academy of Sciences of the USA, 102(36), 12807-12812 (2005)
Development and application of a method for analysis of lufenuron in wheat flour by gas chromatography--mass spectrometry and confirmation of bio-efficacy against Tribolium castaneum (Herbst)(Coleoptera: Tenebrionidae)
Ahire.CK, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 861(1), 16-21 (2008)
Comparison of flea control strategies using imidacloprid or lufenuron on cats in controlled simulated home environment.
EDJ, et al.
American Journal of Veterinary Research, 58(11), 1260-1262 (1997)
Vicente Navarro-Llopis et al.
Pest management science, 66(5), 511-519 (2010-01-27)
The chemosterilisation technique has been demonstrated to reduce the population and fruit damage of the Mediterranean fruit fly, Ceratitis capitata (Wiedemann), in citrus orchards. Field trials showed efficacy by reducing the fruit fly population, which was progressively achieved by continuous
V Navarro-Llopis et al.
Journal of economic entomology, 104(5), 1647-1655 (2011-11-10)
During 2008 and 2009, the efficacy of the combination of two Mediterranean fruit fly, Ceratitis capitata (Wiedemann) (Diptera: Tephritidae), control techniques, sterile insect technique (SIT) and a chemosterilant bait station system (Adress), was tested in three crops: citrus (Citrus spp.)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service