Skip to Content
Merck
All Photos(1)

Key Documents

30540

Supelco

Decane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
Beilstein:
1696981
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

Assay

≥99.8% (GC)

autoignition temp.

410 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

2.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.411 (lit.)
n20/D 1.412

bp

174 °C (lit.)

mp

−30 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decane is an alkane hydrocarbon. It exists as a colourless flammable liquid. It is mostly used as solvent. Its vapors may be narcotic at high concentrations.

Application

Decane may have been used as internal standard in GC analysis for determining the product yield during iron-catalyzed cross coupling performed of primary/secondary alkyl halides with aryl Grignard reagent.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Iron-catalyzed cross-coupling of primary and secondary alkyl halides with aryl grignard reagents.
Nakamura M
Journal of the American Chemical Society, 126(12), 3686-3687 (2004)
Mahvash Karimi et al.
Colloids and surfaces. B, Biointerfaces, 95, 129-136 (2012-03-27)
Wettability alteration is considered to be one of the important mechanisms that lead to increased oil recovery during microbial enhanced oil recovery (MEOR) processes. Changes in wettability will greatly influence the petrophysical properties of the reservoir rocks and determine the
Florent Darriet et al.
Planta medica, 78(4), 386-389 (2012-01-20)
Investigation of the essential oil of the aerial parts of Eryngium maritimum L. from Corsica led to the isolation of one known sesquiterpene (1) and three new oxygenated sesquiterpenes with a muurolane or cadinane skeleton (2-4). Structure assignments of 4
Ilya V Pobelov et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(16), 164210-164210 (2012-04-03)
We describe a new setup for simultaneous measurements of force and current in conductive nanocontacts in a liquid environment with a high sampling rate and resolution. A lab-built current-to-voltage converter allows measurements of the current over seven orders of magnitude.
Ching-Wei Njauw et al.
Langmuir : the ACS journal of surfaces and colloids, 29(12), 3879-3888 (2013-02-28)
It has been known that the addition of bile salts to lecithin organosols induces the formation of reverse wormlike micelles and that the worms are similar to long polymer chains that entangle each other to form viscoelastic solutions. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service