Skip to Content
Merck
All Photos(2)

Key Documents

30269

Supelco

Emodin

analytical standard

Synonym(s):

1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione, 1,3,8-Tri­hydroxy-6-methyl­anthra­quinone, 6-Methyl-1,3,8-tri­hydroxy­anthra­quinone, Archin, Emodol, Frangula-emodin, Frangulic acid, Rheum emodin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
1888141
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(O)c2C(=O)c3c(O)cc(O)cc3C(=O)c2c1

InChI

1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3

InChI key

RHMXXJGYXNZAPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gopal Srinivas et al.
Medicinal research reviews, 27(5), 591-608 (2006-10-05)
Anthraquinones represent a large family of compounds having diverse biological properties. Emodin (1,3,8-trihydroxy-6-methylanthraquinone) is a naturally occurring anthraquinone present in the roots and barks of numerous plants, molds, and lichens, and an active ingredient of various Chinese herbs. Earlier studies
Wonhwa Lee et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 52, 97-104 (2012-11-14)
High mobility group box 1 (HMGB1) protein acts as a potent proinflammatory cytokine and is involved in the pathogenesis of several vascular diseases, such as, systemic vasculitis and sepsis. Emodin-6-O-β-D-glucoside (EG) is a new active compound from Reynoutria japonica, and
Tzong-Der Way et al.
European journal of cancer (Oxford, England : 1990), 50(2), 366-378 (2013-10-26)
Head and neck squamous cell carcinoma (HNSCC) is one of the leading causes of cancer deaths worldwide. In recent studies, a crucial link has been discovered between the acquisition of metastatic traits and tumour-initiating abilities in cancer cells during the
Kejuan Song et al.
Oncology reports, 39(5), 2063-2070 (2018-03-08)
Ovarian cancer is a highly metastatic malignancy and a leading cause of cancer-related death in postmenopausal women. Emodin is a natural anthraquinone isolated from several traditional Chinese medicines including Rhubarb and Polygonum cuspidatum. Recently, emodin was demonstrated to reduce the growth
Jie Yang et al.
Experimental cell research, 319(20), 3182-3189 (2013-10-22)
The activation of nuclear factor-κB (NF-κB) and the subsequent overexpression of its downstream targets transforming growth factor-β1 (TGF-β1) and fibronectin (FN) are among the hallmarks for the progressive diabetic nephropathy. Our previous studies demonstrated that emodin ameliorated renal injury and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service