Skip to Content
Merck
All Photos(1)

Key Documents

27098

Supelco

Chromoionophore VIII

Selectophore

Synonym(s):

3′,3″,5′,5″-Tetrabromophenolphthaleinethyl ester, TBPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14Br4O4
CAS Number:
Molecular Weight:
661.96
Beilstein:
2228548
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

solid

SMILES string

CCOC(=O)c1ccccc1\C(c2cc(Br)c(O)c(Br)c2)=C3/C=C(Br)C(=O)C(Br)=C3

InChI

1S/C22H14Br4O4/c1-2-30-22(29)14-6-4-3-5-13(14)19(11-7-15(23)20(27)16(24)8-11)12-9-17(25)21(28)18(26)10-12/h3-10,27H,2H2,1H3

InChI key

SQFXATUXPUCFFO-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

H+-selective chromoionophore employed in combination with ETH 157 (Cat. No. 71733) for the detn. of ammonia in air

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.J. West et al.
Analytical Chemistry, 64, 533-533 (1992)
Kanokwan Kiwfo et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-09)
A mono-segmented sequential injection lab-at-valve (SI-LAV) system for the determination of albumin, glucose, and creatinine, three key biomarkers in diabetes screening and diagnosis, was developed as a single system for multi-analyte analysis. The mono-segmentation technique was employed for in-line dilution
Spectrophotometric determination of histamine in mast cells, muscle and urine by solvent extraction with copper(II) and tetrabromophenolphthalein ethyl ester.
T Sakai et al.
The Analyst, 109(12), 1569-1572 (1984-12-01)
The application of the TBP test to bile.
E G Saker
Journal of analytical toxicology, 8(1), 51-51 (1984-01-01)
H T Chang et al.
Electrophoresis, 22(3), 394-398 (2001-03-22)
Electroelution of protein bands from a gel has advantages over the competitive common technique requiring gel sectioning with respect to yield, speed and the potential for computer-controlled application to multicomponent two-dimensional (2-D) gels. The electroelution design for the commercial high-performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service