Key Documents
21967
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1100TLL, Kel-F CTFE Luer lock, volume 100 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
material
Kel-F CTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 86020
needle L
, n/a
volume
100 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1100TLL (PTFE Luer lock)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Application
It has been used in syringe pumps to set gas and liquid volumetric flow rate while investigating mechanism which enhances heat transfer model for gas-liquid slug flows.[3] It has also been used in pumping anolyte and catholyte for microbial electrochemical analysis of Geobacter sulfurreducens in response to different chemicals using laminar-flow microfluidic device.[4]
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Heat transfer model for gas-liquid slug flows under constant flux.
Walsh, Patrick A., Edmond J. Walsh, and Yuri S. Muzychka
Int J Heat M T, 53.15, 3193-3201 (2010)
Guy, Jeffrey S.
Pharmacology for the Prehospital Professional null
A Laminar?Flow Microfluidic Device for Quantitative Analysis of Microbial Electrochemical Activity.
Li, Zhongjian, et al.
ChemSusChem, 5.6, 1119-1123 (2012)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)