19399
Nitric acid/Dipicolinic acid concentrate
HNO3 17 mM and Dipicolinic acid 17 mM in water, IC eluent concentrate (10x) for Metrosep C 6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161700
NACRES:
NB.21
Recommended Products
Quality Level
quality
HNO3 17 mM and Dipicolinic acid 17 mM in water
shelf life
limited shelf life, expiry date on the label
concentration
17 mM Dipicolinic acid in H2O
17 mM HNO3 in H2O
technique(s)
ion chromatography: suitable
General description
This eluent concentrate is application tested. Information about the exact contents and expiry date is described in the certificate.
Application
Metrohm IC application note AN-C-173: Cations inroadside snow.
Metrohm IC application note AN-C-145: Ammoniumtraces besides excess sodium using 940 Professional IC Vario and directconductivity detection.
Metrohm IC application note AN-C-142: Separationof the standard cations on the high-capacity Metrosep C 6 separation columns
Metrohm IC application note AN-C-145: Ammoniumtraces besides excess sodium using 940 Professional IC Vario and directconductivity detection.
Metrohm IC application note AN-C-142: Separationof the standard cations on the high-capacity Metrosep C 6 separation columns
Caution
May form precipitation at storage below room temperature.
Linkage
Visit the IC Portal to learn more
Preparation Note
For the preparation of this CRM only materials of highest purity were used.
Analysis Note
HNO3 17 mM and Dipicolinic acid 17 mM in water
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service