17215
WL Differential Agar
suitable for microbiology, NutriSelect® Plus
Synonym(s):
WLD Agar, Wallerstein Differential Agar
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
actidione (cycloheximide), 0.004 g/L
agar, 20 g/L
bromocresol green, 0.022 g/L
calcium chloride, 0.125 g/L
casein enzymic hydrolysate, 5 g/L
dextrose, 50 g/L
ferric chloride, 0.0025 g/L
magnesium sulfate, 0.125 g/L
manganese sulfate, 0.0025 g/L
monopotassium phosphate, 0.55 g/L
potassium chloride, 0.425 g/L
yeast extract, 4 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
5.5±0.2 (25 °C)
application(s)
bioburden testing
food and beverages
microbiology
suitability
selective and differential for Escherichia coli
selective and differential for Lactobacillus spp.
selective and differential for Proteus spp.
selective and differential for bacteria (General Media)
selective and differential for coliforms
Related Categories
General description
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service