Skip to Content
Merck
All Photos(1)

Documents

16957

Supelco

Diethanolamine

analytical standard

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

Assay

≥99.7% (GC)

autoignition temp.

689 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethanolamine (DEA) is an organic chemical compound, which comprises of a secondary amine and a dialcohol. It mainly acts as a weak base. DEA can be used in preparing long-chain fatty acids, namely diethanolamides and diethanolamine salts, which find application in liquid laundry and dishwashing detergents, cosmetics, shampoos, hair conditioners, textile processing, and also in pharmaceutical industries.

Application

Diethanolamine may be used as an analytical reference standard for the determination of the analyte in air samples, aqueous solutions, and shampoo products by various chromatographic techniques.
It was used as reference standard for determination of DEA in air using high-performance liquid chromatograph equipped with a fluorescence detector or photo-diode array detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic detection of diethanolamine and its degradation products
Choy, Edward T and Meisen, Axel
Journal of Chromatography A, 187(1), 145-152 (1980)
Determination method for mono-and diethanolamine in workplace air by high-performance liquid chromatography
Takeuchi A, et al.
Journal of Occupational Health, 187(1), 12-0064 (2012)
Dip-coating of TiO2 films using a sol derived from Ti (Oi-Pr) 4-diethanolamine-H2O-i-PrOH system.
Takahashi, Yasutaka, and Yoshihiro Matsuoka.
J. Mater. Sci., 23.6, 2259-2266 (1988)
Determination of diethanolamine in shampoo products containing fatty acid diethanolamides by liquid chromatography with a thermal energy analyzer
Chou HJ
Journal of AOAC (Association of Official Analytical Chemists) International, 88(2), 592-594 (2005)
Determination of monoethanolamine and diethanolamine in air
Levin JO, et al.
Annals of Occupational Hygiene, 33(2), 175-180 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service