Skip to Content
Merck
All Photos(1)

Key Documents

16558

Supelco

Fragrance allergen mix A2

40 components in tert-Butyl methyl ether, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

quality

40 components in tert-Butyl methyl ether

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
This product in combination with fragrance allergen mix A1 is in compliance with the IFRA method for quantification of 57 allergen fragrances.

Application

ApplicationNote: "Allergenic Fragrance Testing – New Certified Reference Materialsand GC-FID/GC-MS Application" article from AnalytixReporter issue 3

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    3-Propylidenephthalide
    α-Acetyl cedrene
    Isoeugenyl acetate
    α-Amyl-trans-cinnamaldehyde
    Amyl salicylate
    Benzaldehyde
    Benzyl benzoate
    Benzyl cinnamate
    Benzyl salicylate
    Butylphenyl methylpropional
    D-Camphor
    (+)-Carvone
    trans-Cinnamaldehyde
    Citral total
    Coumarin
    Damascenone
    α-Damascone
    (E)-β-Damascone, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
    (E)-β-Damascone
    δ-Damascone
    Dimethylbenzylcarbinyl acetate (DMBCA acetate)
    Eugenyl acetate
    Galaxolide 1 (Hexamethylindanopyran)
    Galaxolide 2(Hexamethylindanopyran)
    Geranyl acetate
    Hexadecanolactone / Dihydroambrettolide
    α-Hexylcinnamaldehyde
    Hydroxycitronellal
    Isomethylionone alpha
    Linalyl acetate
    Hydroxyisohexyl 3-cyclohexene carboxaldehyde
    Methyl salicylate
    Folione (Methyl 2-octynoate)
    Salicylaldehyde
    Terpinolene
    Vanillin
    ISO E alpha + beta+ gamma
    See All (37)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service