Skip to Content
Merck
All Photos(1)

Key Documents

15222

Supelco

N,O-Bis(trimethylsilyl)trifluoroacetamide

for GC derivatization, LiChropur, ≥99.0%

Synonym(s):

BSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥99.0% (GC)
≥99.0%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.384 (lit.)
n20/D 1.384

bp

45-50 °C/14 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+

InChI key

XCOBLONWWXQEBS-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,O-Bis(trimethylsilyl)trifluoroacetamide (BSTFA) is an excellent silylation reagent. For some sterically hindered primary or secondary nitro compounds, N,O-bis(trimethylsilyl)acetamide and BSTFA achieve better results than normal silylation conditions. BSTFA finds application in derivatizing amines. It is suitable for silyaltion of carboxylic acids, phenols, ureas, amides and alcohols.

Application

Learn more in the Product Information
BSTFA + 1% TMCS (trimethylsilyl) in ethyl acetate, acetonitrile and dichloromethane solvents may be used for silylation of steroid hormone 17a-ethinylestradiol (EE2) using gas chromatography-mass spectrometry (GC-MS). It may also be used for silylation of endogenous 2-monoglyceride, present in canine gut that binds to cannabinoid receptors.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F

Flash Point(C)

34 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Modern Methods of Pharmaceutical Analysis.
Schirmer RE.
Science, 2, 215-216 (1990)
R Mechoulam et al.
Biochemical pharmacology, 50(1), 83-90 (1995-06-29)
In this study, we report the isolation from canine intestines of 2-arachidonyl glycerol (2-Ara-Gl). Its structure was determined by mass spectrometry and by direct comparison with a synthetic sample. 2-Ara-Gl bound to membranes from cells transiently transfected with expression plasmids
Bo Wang et al.
Foods (Basel, Switzerland), 9(5) (2020-05-24)
A method based on accelerated solvent extraction (ASE) coupled with gas chromatography tandem mass spectrometry (GC-MS/MS) was developed for the quantitative analysis of spectinomycin and lincomycin in poultry egg (whole egg, albumen and yolk) samples. In this work, the samples
Greene's Protective Groups in Organic Synthesis.
Science (2014)
Houben-Weyl Methods of Molecular Transformations.
Aggarwal VK
Science of Synthesis Knowledge Updates, 27, 593-594 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service