14379
MS calibration solution
API; sodium iodide (2 μg/μL) and cesium iodide (50 ng/μL) in 50/50 2-propanol/water
Synonym(s):
Mass spectrometry calibration standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
NA.21
Recommended Products
Quality Level
form
liquid
density
0.910-0.950 g/mL at 20 °C
suitability
in accordance for LC-MS
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
71.6 °F
Flash Point(C)
22 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Heeseon An et al.
Molecular cell, 74(5), 891-908 (2019-04-23)
Cells respond to nutrient stress by trafficking cytosolic contents to lysosomes for degradation via macroautophagy. The endoplasmic reticulum (ER) serves as an initiation site for autophagosomes and is also remodeled in response to nutrient stress through ER-phagy, a form of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service