Skip to Content
Merck
All Photos(3)

Key Documents

141194

Sigma-Aldrich

Ponceau S

Dye content 75 %

Synonym(s):

3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
Colour Index Number:
27195
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 75%

technique(s)

microbe id | staining: suitable

λmax

352 nm
520 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-8-3-14(11-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,27H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23+,26-25+;;;;

InChI key

KQHKSGRIBYJYFX-WNDMICSESA-J

Looking for similar products? Visit Product Comparison Guide

General description

Ponceau S (C.l. 27195) is an anionic, very hydrophilic disazo dye. It is also known as fast ponceau 2B.

Application

Ponceau S is widely used to stain proteins on gel electropherograms. It is a suitable stain for proteins adsorbed onto nitrocellulose membranes. It also has applications in botanical staining manuals as a stain for sieve plate elements.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Internal amino acid sequence analysis of proteins separated by one- or two-dimensional gel electrophoresis after in situ protease digestion on nitrocellulose.
Aebersold RH
Proceedings of the National Academy of Sciences of the USA, 84(20), 6970-6974 (1987)
An intramembrane modulator of the ErbB2 receptor tyrosine kinase that potentiates neuregulin signaling.
Carraway KL III
The Journal of Biological Chemistry, 274(9), 5263-5266 (1999)
Expression of cyclooxygenase-2 (COX-2) in human invasive transitional cell carcinoma (TCC) of the urinary bladder.
Mohammed SI
Cancer Research, 59(22), 5647-5650 (1999)
Sarah Nicklas et al.
PloS one, 7(1), e30445-e30445 (2012-02-03)
Limb girdle muscular dystrophy type 2H (LGMD2H) is an inherited autosomal recessive disease of skeletal muscle caused by a mutation in the TRIM32 gene. Currently its pathogenesis is entirely unclear. Typically the regeneration process of adult skeletal muscle during growth
T G van Kooten et al.
Journal of biomedical materials research, 54(3), 385-389 (2001-02-24)
Research of cell-biomaterial interactions is building on knowledge and methods available in cell and molecular biology. Western blotting is one of the options to characterize protein expression in cell populations. Method transfer to biomaterial model systems is not trivial because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service