Skip to Content
Merck
All Photos(1)

Documents

14023

Sigma-Aldrich

Titanium(IV) oxysulfate

≥29% Ti (as TiO2) basis, technical

Synonym(s):

Titanyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiOSO4 · aq
CAS Number:
Molecular Weight:
159.93 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

technical

form

solid

reaction suitability

core: titanium
reagent type: catalyst

concentration

≥29% (Ti (as TiO2))

impurities

sulfonic acid
≤17% free acid (as H2SO4)

anion traces

chloride (Cl-): ≤500 mg/kg

cation traces

Fe: ≤500 mg/kg

SMILES string

O.O=[Ti++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.H2O.O.Ti/c1-5(2,3)4;;;/h(H2,1,2,3,4);1H2;;/q;;;+2/p-2

InChI key

SWNAGNPMPHSIJF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Titanium(IV) oxysulfate has been used as a precursor to synthesize a wide range of titanium dioxide (TiO2) photocatalysts.
It can also be used in the synthesis of freeze dried titania foams, and WO3-loaded TiO2 photocatalyst applicable for the oxidation of trans-ferulic acid to vanillin.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Wende et al.
Mutation research. Genetic toxicology and environmental mutagenesis, 798-799, 48-54 (2016-03-21)
Cold atmospheric pressure plasmas represent a favorable option for the treatment of heat sensitive materials and human or animal tissue. Beneficial effects have been documented in a variety of medical conditions, e.g., in the treatment of chronic wounds. It is
Karoly Kozma et al.
Dalton transactions (Cambridge, England : 2003), 48(29), 11086-11093 (2019-07-02)
TiO2 is manufactured for white pigments, solar cells, self-cleaning surfaces and devices, and other photocatalytic applications. The industrial synthesis of TiO2 entails: (1) the dissolution of ilmenite ore (FeTiO3) in aqueous sulfuric acid which precipitates the Fe while retaining the
Stanislav S Fedotov et al.
Nature communications, 11(1), 1484-1484 (2020-03-22)
The rapid progress in mass-market applications of metal-ion batteries intensifies the development of economically feasible electrode materials based on earth-abundant elements. Here, we report on a record-breaking titanium-based positive electrode material, KTiPO4F, exhibiting a superior electrode potential of 3.6 V in
Highly photoactive 2D titanium dioxide nanostructures prepared from lyophilized aqueous colloids of peroxo-polytitanic acid
Subrt J, et al.
Materials Research Bulletin, 49, 405-412 (2014)
Photocatalytic green synthesis of piperonal in aqueous TiO2 suspension
Bellardita M, et al.
Applied Catalysis. B, Environmental, 144, 607-613 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service