Skip to Content
Merck
All Photos(1)

Documents

10981

Supelco

Polyvinyl alcohol mounting medium with DABCO®, antifading

pH 8.7

Synonym(s):

PVA-DABCO®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NA.25

refractive index

n20/D 1.377

Quality Level

pH

8.7

density

1.08 g/mL at 20 °C

suitability

in accordance for application

shipped in

wet ice

storage temp.

2-8°C

Physical form

Aqueous solution containing tris(hydroxymethyl)aminomethane, tris(hydroxymethyl)aminomethane hydrochloride, polyvinyl alcohol 22000, glycerol, and 1,4-diazabicyclo[2.2.2]octane

Legal Information

DABCO is a registered trademark of Air Products & Chemicals, Inc.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sierra Simpson et al.
eNeuro, 7(3) (2020-04-29)
Substance use disorders have a complex etiology. Genetics, the environment, and behavior all play a role in the initiation, escalation, and relapse of drug use. Recently, opioid use disorder has become a national health crisis. One aspect of opioid addiction
Laisel Martinez et al.
Journal of immunology (Baltimore, Md. : 1950), 205(9), 2545-2553 (2020-09-18)
Pharmacological activation of integrin CD11b/CD18 (αMβ2, Mac-1, and CR3) shows anti-inflammatory benefits in a variety of animal models of human disease, and it is a novel therapeutic strategy. Reasoning that genetic models can provide an orthogonal and direct system for
Alejandra Rodriguez Celin et al.
PloS one, 10(1), e0116343-e0116343 (2015-01-31)
The developing chick optic tectum is a widely used model of corticogenesis and angiogenesis. Cell behaviors involved in corticogenesis and angiogenesis share several regulatory mechanisms. In this way the 3D organizations of both systems adapt to each other. The consensus
Zurine De Miguel et al.
Molecular psychiatry, 24(8), 1178-1188 (2018-01-10)
Chronic stress is a recognized risk factor for psychiatric and psychological disorders and a potent modulator of adult neurogenesis. Numerous studies have shown that during stress, neurogenesis decreases; however, during the recovery from the stress, neurogenesis increases. Despite the increased
Burkhard H A von Rahden et al.
Journal of experimental & clinical cancer research : CR, 30, 23-23 (2011-02-25)
Investigation of the expression of an intestinal stem cell marker in esophageal adenocarcinomas (EAC) with and without Barrett's Esophagus (BE), with respect to a cancer stem cell (CSC) hypothesis. Expression of a putative intestinal stem cell marker LgR5 was analyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service