Skip to Content
Merck
All Photos(1)

Key Documents

09898

Sigma-Aldrich

Ammonium oxalate monohydrate

BioUltra, ≥99.5% (RT)

Synonym(s):

Oxalic acid diammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
142.11
Beilstein:
3759830
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
solubility:
H2O: 0.1 M at 20 °C, clear, colorless

grade

reagent grade

Quality Level

product line

BioUltra

Assay

≥99.5% (RT)

impurities

insoluble matter, passes filter test

loss

≤0.02% loss on ignition

pH

6.0-7.0 (25 °C, 0.1 M in H2O)

mp

133 °C (dec.) (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 267 nm in H2O at 0.1 M

SMILES string

OC(C(O)=O)=O.N.O.N

InChI

1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2

InChI key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium oxalate monohydrate can be used to study chelators, biological buffers and biochemicals and reagents. Ammonium oxalate monohydrate has been used in an analysis of the effective segregation coefficient keff of different impurities, which suggested that the dehydration energies of cations mainly determine the capability of capture of impurity species by the growing crystal.

Other Notes

Complexes with Cu, thus inhibits spinach chloroplast phenolase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EPR of Mn2+-doped ammonium oxalate monohydrate
Misra, S.K., et al.
Solid State Communications, 59, 409-413 (1986)
Study of segregation coefficient of cationic impurities in ammonium oxalate monohydrate crystals during growth from aqueous solutions
Sangwal, K., et al.
Journal of Crystal Growth, 244, 183-193 (2002)
Study of segregation coefficient of Mn(II) impurity in ammonium oxalate monohydrate crystals and the relationship between segregation coefficient and growth kinetics
Sangwal, K. and Mielniczek-Brzoska, E.
Journal of Crystal Growth, 257, 185-198 (2003)
M. Sato
Phytochemistry, 19, 1939-1939 (1980)
Electrochemiluminescence as a tool for microscopy at the nanoscale.
Lee A, et al.
Proceedings of SPIE, 13, 13-20 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service