Skip to Content
Merck
All Photos(2)

Documents

06709

Supelco

Pinoresinol

analytical standard

Synonym(s):

(+)-Pinoresinol, 4,4′-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol), 4,4′-[(1S,3aR,4S,6aR)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2-methoxyphenol)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

COc1cc(ccc1O)[C@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4ccc(O)c(OC)c4

InChI

1S/C20H22O6/c1-23-17-7-11(3-5-15(17)21)19-13-9-26-20(14(13)10-25-19)12-4-6-16(22)18(8-12)24-2/h3-8,13-14,19-22H,9-10H2,1-2H3/t13-,14-,19+,20+/m0/s1

InChI key

HGXBRUKMWQGOIE-AFHBHXEDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pinoresinol is present in a number of plants including medical plants like Sambucus williamsii, Eucommia ulmoides (duzhong), Styrax sp, Forsythia suspensa, and in extra virgin olive oil. The genus Sambucus is widely distributed in Europe, Asia, and North Africa, and has been used in traditional medicine as an analgesic, antivirus, antiinflammatory, homoeostatic, and diuretic drugs which act on bruises, fractures, and edema. Pinoresinol displays potent antifungal properties by causing damage to the fungal plasma membrane. It exerts anti-oxidative and anti-inflammatory activities by inhibition of TNF- α production (presumably through inhibition of NF- κB and AP-1).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Jie Li et al.
Molecules (Basel, Switzerland), 17(8), 8773-8781 (2012-07-27)
Two new compounds, (2S,3R)-methyl 7-hydroxy-2-(4-hydroxy-3-methoxy-phenyl)-3-(hydroxymethyl)-2,3-dihydrobenzofuran-5-carboxylate (1) and (4R,5S)-5-(3-hydroxy-2,6-dimethylphenyl)-4-isopropyldihydrofuran-2-one (2), tentatively named norcurlignan and limlactone, respectively, were isolated from Liriope muscari, together with the known compound (-)-pinoresinol (3). The structures of these compounds were elucidated and characterized on the basis of
Sullivan Renouard et al.
Planta, 235(1), 85-98 (2011-08-13)
Secoisolariciresinol diglucoside (SDG), the main phytoestrogenic lignan of Linum usitatissimum, is accumulated in the seed coat of flax during its development and pinoresinol-lariciresinol reductase (PLR) is a key enzyme in flax for its synthesis. The promoter of LuPLR1, a flax
J L Peñalvo et al.
European journal of clinical nutrition, 66(7), 795-798 (2012-05-17)
Lignan-rich diets have been associated with favorable health effects through improved metabolic profile. In this study, we hypothesized that dietary lignan intake could be also associated with childhood obesity. We studied prevalent obesity in relation to lignan intake within the
An enantiocomplementary dirigent protein for the enantioselective laccase-catalyzed oxidative coupling of phenols.
Benjamin Pickel et al.
Angewandte Chemie (International ed. in English), 49(1), 202-204 (2009-12-01)
Sheng-Zhuo Huang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 38(1), 64-69 (2013-04-20)
To study the chemical constituents from Daphne acutiloba. The constituents were separated by column chromatography, and their structures were elucidated by spectroscopic data analyses. Fifteen compounds were isolated from the EtOAc extract and identified as wikstroelide M (1), vesiculosin (2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service