Skip to Content
Merck
All Photos(2)

Documents

06679

Sigma-Aldrich

Methoxypolyethylene glycol amine

5,000, extent of labeling: ≥0.17 mmol/g NH2 loading

Synonym(s):

O-(2-Aminoethyl)-O′-methylpolyethylene glycol, Aminopolyethylene glycol monomethyl ether, Methoxypolyoxyethylene amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2(OCH2CH2)nOCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

extent of labeling

≥0.17 mmol/g NH2 loading

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methoxypolyethylene glycol amine is generally used for the chemical modification of biologically active compounds and therapeutic agents. It has been used to increase the aqueous solubility of long-wavelength voltage-sensitive dyes and to prepare the PEG-coated fluorescent beads in studying large-scale rotational cytoplasmic flow.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatial confinement of active microtubule networks induces large-scale rotational cytoplasmic flow
Suzuki K, et al.
Test, 114(11), 2922-2927 (2017)
Tools for Drug Discovery
Aldrich Chemfiles, 5(12), 19-20 (2005)
Enhanced aqueous solubility of long wavelength voltage-sensitive dyes by covalent attachment of polyethylene glycol
Patrick MJ, et al.
Organic & Biomolecular Chemistry, 5(20), 3347-3353 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service