04447
HybriScan™D Waste Water Microthrix parvicella
suitable for microbiology
Synonym(s):
FastScan Waste Water Microthrix parvicella, Waste Water Microthrix parvicella HybriScan™D
About This Item
Recommended Products
technique(s)
microbe id | in situ hybriziation: suitable
Quality Level
application(s)
clinical testing
food and beverages
microbiology
storage temp.
2-8°C
suitability
Microthrix parvicella
General description
Linkage
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service