Skip to Content
Merck
All Photos(1)

Key Documents

02562

Supelco

1,2-Dichloroethane

analytical standard

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39050106
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

Assay

≥99.9% (GC)

autoignition temp.

775 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

15.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.444 (lit.)
n20/D 1.444

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloroethane (Ethylene Dichloride) is a clear, colorless, oily, synthetic, flammable liquid chlorinatied hydrocarbon with a pleasant chloroform-like smell that emits toxic fumes of hydrochloric acid when heated to decomposition. It is primarily used to produce vinyl chloride, and as a solvent in food processing. It can be used in the manufacturing of acetyl cellulose and ethylenediamine. It also can serve as a solvent for oils, fats, waxes, resins, gums,etc.

Application

1,2-Dichloroethane may be used as an analytical reference standard for the determination of the analyte in biological samples, wastewater, and sewage samples by gas chromatography (GC) based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of unknown organic pollutants in sewage by solid-phase extraction combined with gas chromatography-mass spectrometry
Gao H, et al.
Journal of Chromatographic Science, 42(2), 91-99 (2004)
Groundwater Chemicals Desk Reference, December 12, 348-350 (2010)
Yijing Li et al.
Molecular cancer, 11, 87-87 (2012-11-24)
IL-1β is a pleiotropic pro-inflammatory cytokine and its up-regulation is closely associated with various cancers including gastrointestinal tumors. However, it remains unclear how IL-1β may contribute to the initiation and development of these inflammation-associated cancers. Here we investigated the role
E Oikonomou et al.
British journal of cancer, 97(1), 73-84 (2007-06-07)
Most data on the therapeutic potential of tumour necrosis factor-related apoptosis-inducing ligand (TRAIL) as well as resistance to FAS ligand (FASL) in colorectal cancer have come from in vitro studies using cell lines. To gain a clearer understanding about the
Development of a procedure for the gas-chromatographic determination of vinyl chloride and 1, 2-dichloroethane in blood with the use of headspace analysis
Alekseenko AN, et al.
Journal of Analytical Chemistry, 65(7), 739-742 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service