Skip to Content
Merck
All Photos(1)

Documents

40-2023

Sigma-Aldrich

Bromothymol Blue solution

pH 6.0-7.6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28Br2O5S
CAS Number:
Molecular Weight:
624.38
Beilstein:
373934
MDL number:
UNSPSC Code:
12171502
PubChem Substance ID:

form

liquid

availability

available only in Japan

pH

6.0-7.6

SMILES string

CC(C)c1cc(c(C)c(Br)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c(O)c(Br)c4C

InChI

1S/C27H28Br2O5S/c1-13(2)17-11-20(15(5)23(28)25(17)30)27(19-9-7-8-10-22(19)35(32,33)34-27)21-12-18(14(3)4)26(31)24(29)16(21)6/h7-14,30-31H,1-6H3

InChI key

NUHCTOLBWMJMLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

pH indicator

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana A Sanchez et al.
Analytica chimica acta, 694(1-2), 95-99 (2011-05-14)
Liquid-liquid microextraction without phase segmentation was implemented in a multicommuted flow system for determination of the anti-hypertensive diltiazem. The procedure was based on ion pair formation between the drug and the dye bromothymol blue at pH 3.5. The detection was
Kristina Jurcic et al.
Journal of chromatography. A, 1134(1-2), 317-325 (2006-10-07)
An effective sample preconcentration technique for proteins and peptides was recently developed using capillary electrophoresis (CE) with discontinuous buffers [C.A. Nesbitt, J.T.-M. Lo, K.K.-C. Yeung, J. Chromatogr. A 1073 (2005) 175]. Two buffers of different pH created a junction to
Bambang Kuswandi et al.
Talanta, 74(4), 613-618 (2008-03-29)
An optical fiber biosensor consisting of acetylcholinesterase (AChE) and bromothymol blue (BTB) doped sol-gel film was employed to detect organophosphate pesticide chlorpyrifos. The main advantage of this optical biosensor is the use of a single sol-gel film with immobilized AChE
K R Klein et al.
Journal of clinical microbiology, 47(11), 3669-3672 (2009-10-02)
Cryptococcus neoformans and Cryptococcus gattii are closely related pathogenic fungi. Cryptococcus neoformans is ecologically widespread and affects primarily immunocompromised patients, while C. gattii is traditionally found in tropical climates and has been reported to cause disease in immunocompetent patients. l-Canavanine
Sevgi Tatar Ulu
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(3-4), 778-783 (2006-10-13)
A simple, rapid, accurate, precise and sensitive colorimetric method for the determination of finasteride in tablets is described. The proposed methods are based on the formation of ion-pair complexes between the examined drug with bromophenol blue (BPB), bromocresol green (BCG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service