Skip to Content
Merck
All Photos(1)

Key Documents

30-4160

Sigma-Aldrich

Triethanolamine

JIS special grade, ≥98.0%

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein:
1699263
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
JIS special grade
bp:
190-193 °C/5 mmHg (lit.)
vapor pressure:
0.01 mmHg ( 20 °C)

grade

JIS special grade

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.485 (lit.)

pH

10.5-11.5 (25 °C, 149 g/L)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F

Flash Point(C)

179 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liviu Ungur et al.
Journal of the American Chemical Society, 134(45), 18554-18557 (2012-11-01)
A toroidal magnetic moment in the absence of conventional total magnetic moment is observed in a {Dy(6)} ring. The reason for the net toroidal arrangement of the local magnetic moments is the high symmetry of the complex in combination with
Stuart K Langley et al.
Dalton transactions (Cambridge, England : 2003), 41(32), 9789-9796 (2012-07-11)
The synthesis, structures and magnetic properties of two new mixed-valence heptanuclear manganese clusters are described. Both complexes utilize triethanolamine (teaH(3)) as a bridging ligand, displaying near planar, disc-like metal topologies and are of formulae [Mn(II)(4)Mn(IV)(3)(tea)(teaH(2))(3)(peolH)(4)](BF(4))(2)·solv (1) and [Mn(II)(4)Mn(III)(3)F(3)(tea)(teaH)(teaH(2))(2)(piv)(4)(Hpiv)(chp)(3)]·0.5MeCN (2). Compound
Xiaoxuan Liu et al.
Bioconjugate chemistry, 22(12), 2461-2473 (2011-11-08)
With the aim of developing dendrimer nanovectors with a precisely controlled architecture and flexible structure for DNA transfection, we designed PAMAM dendrimers bearing a triethanolamine (TEA) core, with branching units pointing away from the center to create void spaces, reduce
Xiaoxuan Liu et al.
Molecular pharmaceutics, 9(3), 470-481 (2012-01-03)
Successful achievement of RNA interference in therapeutic applications requires safe and efficient vectors for siRNA delivery. In the present study, we demonstrate that a triethanolamine (TEA)-core PAMAM dendrimer of generation 5 (G(5)) is able to deliver sticky siRNAs bearing complementary
Imad J Matouk et al.
Biochimica et biophysica acta, 1843(7), 1414-1426 (2014-04-08)
The oncofetal H19 gene transcribes a long non-coding RNA(lncRNA) that is essential for tumor growth. Here we found that numerous established inducers of epithelial to mesenchymal transition(EMT) also induced H19/miR-675 expression. Both TGF-β and hypoxia concomitantly induced H19 and miR-675

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service