Skip to Content
Merck
All Photos(1)

Key Documents

28-4330

Sigma-Aldrich

Sodium sulfite

JIS special grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥97.0%
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

≥97.0%

form

solid

availability

available only in Japan

pH

9.0-10.5 (25 °C, 126 g/L)

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Comparative Studies of Organic Contaminant Removal in Different Calcium Sulfite-Enhanced Oxidant/Fe(II) Systems: This research explores the effectiveness of sodium sulfite in enhancing the removal of organic contaminants through various oxidant systems. (Zhou et al., 2023).

  • Review of Traditional and Emerging Residual Chlorine Quenchers on Disinfection By-Products: Focuses on the role of sodium sulfite as a residual chlorine quencher in water treatment processes, analyzing its impact on reducing harmful by-products and enhancing water quality (Li et al., 2023).

  • Boosting the Photoelectrochemical Performance of Bismuth Vanadate Photoanode Through Homojunction Construction: Investigates how sodium sulfite can improve the photoelectrochemical properties of bismuth vanadate, significant for renewable energy applications, particularly in solar energy conversion (Wang et al., 2023).

  • Light-driven Oxidative Demethylation Reaction Catalyzed by a Rieske-type Non-heme Iron Enzyme Stc2: Demonstrates the use of sodium sulfite in enzymatic reactions to achieve oxidative demethylation, which is crucial for synthetic biology and enzyme engineering (Hu et al., 2022).

  • Preparation of Nano Disperse Dyes Using Sulfomethylated Lignin: Effects of Sulfonic Group Contents: Highlights the application of sodium sulfite in the preparation of environmentally friendly nano disperse dyes from lignin, emphasizing its potential in sustainable material science (Tang et al., 2023).

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Michelle Cilia et al.
PloS one, 7(10), e48177-e48177 (2012-11-03)
Circulative transmission of viruses in the Luteoviridae, such as cereal yellow dwarf virus (CYDV), requires a series of precisely orchestrated interactions between virus, plant, and aphid proteins. Natural selection has favored these viruses to be retained in the phloem to
Qiang Yang et al.
Bioresource technology, 126, 336-340 (2012-11-06)
Agave americana is one of commonly grown agave species but currently less valuable because its large flower stalk cannot be used for producing alcoholic beverage. In the present study, the stalk was pretreated with dilute acid (DA), sulfite (SPORL), and
Jingang Wang et al.
Chemosphere, 91(3), 351-357 (2012-12-26)
This paper presents a simple, easy and reliable method for determination of EDTA (ethylenediaminetetraacetic acid) in aqueous system. Using EDTA chelating with ferric irons, an excessive amount of Fe(3+) was added to EDTA solution, and Fe(3+) in excess was reduced
Fabian Leinisch et al.
Chemical research in toxicology, 24(12), 2217-2226 (2011-10-19)
Nitrone spin traps such as 5,5-dimethyl-1-pyrroline-N-oxide (DMPO) are commonly used for free radical detection. Though proven examples are rare, artifact formation must be considered. For example, the Forrester-Hepburn mechanism yields the same radical adduct as that formed by genuine radical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service