Skip to Content
Merck
All Photos(1)

Key Documents

23-0120

Sigma-Aldrich

1-Octanol

JIS special grade, ≥98.0%

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
JIS special grade
technique(s):
HPLC: suitable
bp:
196 °C (lit.)
vapor pressure:
0.14 mmHg ( 25 °C)

grade

JIS special grade

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

523 °F

expl. lim.

8 % (lit.)

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

177.8 °F

Flash Point(C)

81 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashraf S Al-Madhoun et al.
Cancer research, 64(17), 6280-6286 (2004-09-03)
Thymidine analogs containing o-carboranylalkyl groups at the 3-position were screened as potential substrates for human thymidine kinase 1 (TK1), an enzyme that is selectively expressed in a variety of rapidly proliferating cells, including tumor cells. On the basis of previous
R O Potts et al.
Pharmaceutical research, 9(5), 663-669 (1992-05-01)
Published permeability coefficient (Kp) data for the transport of a large group of compounds through mammalian epidermis were analyzed by a simple model based upon permeant size [molecular volume (MV) or molecular weight (MW)] and octanol/water partition coefficient (Koct). The
Sophie Martel et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(1-2), 21-29 (2012-11-08)
Lipophilicity is a crucial parameter in drug development since it impacts both ADME properties and target affinity of drug candidates. In early drug discovery stage, accurate tools for logP prediction are highly desired. Many calculation methods were developed to aid
J Franco et al.
Acta biomaterialia, 6(1), 218-228 (2009-07-01)
The development of materials to support bone regeneration requires flexible fabrication technologies able to tailor chemistry and architecture for specific applications. In this work we describe the preparation of ceramic-based inks for robotic-assisted deposition (robocasting) using Pluronic F-127 solutions. This
H Okazawa et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 35(12), 1910-1915 (1994-12-01)
Copper-62-pyruvaldehyde bis(N4-methylthiosemicarbazone) copper II (62Cu-PTSM) has been proposed as a generator-produced positron-emitting tracer for perfusion imaging. To evaluate the characteristics of 62Cu-PTSM as a cerebral perfusion tracer, brain PET images of 62Cu-PTSM were compared with cerebral blood flow (CBF). Following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service