Skip to Content
Merck
All Photos(1)

Key Documents

19-3540

Sigma-Aldrich

4-Methyl-2-pentanone

JIS special grade, ≥99.5%

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
605399
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

grade

JIS special grade

vapor density

3.5 (vs air)

vapor pressure

15.7 mmHg

Assay

≥99.5%

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

availability

available only in Japan

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginette Truchon et al.
Journal of occupational and environmental hygiene, 3(3), 137-143 (2006-02-09)
Compartmental and physiologically based toxicokinetic modeling coupled with Monte Carlo simulation were used to quantify the impact of biological variability (physiological, biochemical, and anatomic parameters) on the values of a series of bio-indicators of metal and organic industrial chemical exposures.
H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Serife Saçmaci et al.
Analytica chimica acta, 623(1), 46-52 (2008-07-10)
A method for speciation, preconcentration and separation of Fe(II) and Fe(III) in different matrices was developed using solvent extraction and flame atomic absorption spectrometry. 4-Acetyl-5-methyl-1-phenyl-1H-pyrazole-3-carboxylic acid (AMPC) was used as a new complexing reagent for Fe(III). The Fe(III)-AMPC complex was
Matthew D Stout et al.
Toxicology, 244(2-3), 209-219 (2008-01-08)
Methyl isobutyl ketone (MIBK) is primarily used as a denaturant for rubbing alcohol, as a solvent and in the manufacture of methyl amyl alcohol. Inhalation of vapors is the most likely route of exposure in the work place. In order
Kieron J Doick et al.
Environmental science & technology, 39(17), 6575-6583 (2005-09-30)
Organic matter is considered to be the single most importantfactor limiting availability and mobility of persistent organic pollutants (POPs) in soil. This study aimed to characterize the distribution of 14C-PCB (congeners 28 and 52) and 14C-PAH (fluoranthene and benzo[a]pyrene) residues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service