Skip to Content
Merck
All Photos(1)

Key Documents

13-0190

Sigma-Aldrich

Hematoxylin monohydrate

SAJ special grade

Synonym(s):

Hematoxylin, Natural Black 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O6 · xH2O
CAS Number:
Molecular Weight:
302.28 (anhydrous basis)
Colour Index Number:
75290
Beilstein:
91400
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

SAJ special grade

form

powder

availability

available only in Japan

mp

200 °C (dec.) (lit.)

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinchita Roy Chowdhuri et al.
Acta cytologica, 56(6), 622-631 (2012-12-05)
In the past decade molecular diagnostics has changed the clinical management of lung adenocarcinoma patients. Molecular diagnostics, however, is largely dependent on the quantity and quality of the tumor DNA that is retrieved from the tissue or cytology samples. Frequently
Biswanath Maity et al.
Methods in cell biology, 113, 81-105 (2013-01-16)
The purpose of this protocol is to describe various methodologies used to detect the distribution and localization of specific proteins within individual cells or tissues using immunostaining, defined as the use of specific antibodies to detect a single target protein.
Sait Sen et al.
Turk patoloji dergisi, 28(3), 204-212 (2012-09-27)
Systemic amyloidosis is a rare disorder, characterized by extracellular accumulation of Congo red positive fibrillar amyloid protein deposits that have an amorphous, eosinophilic appearance on hematoxylin-eosin stained preparations. The kidney is the most commonly affected organ by systemic amyloidosis. Congo
Douglas A Hirth et al.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society, 20(6), 918-927 (2012-11-07)
Histopathology remains the gold standard for evaluation of burn depth, progression, and healing, but burn literature offers little guidance on the best stains for analysis of these complex and evolving injuries. A battery of histochemical and immunohistochemical stains was compared
Makoto Miyagawa
Rinsho byori. The Japanese journal of clinical pathology, 61(4), 313-317 (2013-07-17)
The present study was undertaken to establish the experimental animal model by histological staining methods for minerals. After intraperitoneal injections of minerals, precipitates deposited on the surface of the liver. Liver tissues were fixed in paraformaldehyde, embedded in paraffin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service