Skip to Content
Merck
All Photos(1)

Key Documents

09-0240

Sigma-Aldrich

Eosin Y

SAJ special grade, ≥85.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
Colour Index Number:
45380
Beilstein:
3586809
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

SAJ special grade

Assay

≥85.0%

availability

available only in Japan

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C20H8Br4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

SEACYXSIPDVVMV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

376.9 °F - Pensky-Martens closed cup

Flash Point(C)

191.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Shih et al.
Macromolecular rapid communications, 34(3), 269-273 (2013-02-07)
The utility of visible-light-mediated polymerization in tissue engineering has been limited due to the necessary use of potentially cytotoxic coinitiator and comonomer. Here, we report a visible-light-mediated thiol-ene hydrogelation scheme using eosin-Y as the only photoinitiator. Under visible light exposure
Han Shih et al.
ACS applied materials & interfaces, 5(5), 1673-1680 (2013-02-07)
Interfacial visible light-mediated thiol-ene photoclick reactions were developed for preparing step-growth hydrogels with multilayer structures. The effect of a noncleavage type photoinitiator eosin-Y on visible-light-mediated thiol-ene photopolymerization was first characterized using in situ photorheometry, gel fraction, and equilibrium swelling ratio.
Toshihisa Yashiro et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(48), 17212-17217 (2014-11-19)
Males and females are in conflict over genetic transmission in the evolution of parthenogenesis, because it enhances female reproductive output but deprives the males' genetic contribution. For males, any trait that coerces females into sexual reproduction should increase their fitness.
Patricia S Nielsen et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 26(3), 404-413 (2012-11-24)
Cellular proliferation is correlated with the progression of melanoma. Accordingly, the proliferation index of H&E-stained thin melanomas was recently included in the staging system of the American Joint Committee on Cancer. Yet, the immunohistochemical markers of proliferation phosphohistone H3 and
Jie Du et al.
Fish & shellfish immunology, 34(1), 315-323 (2012-11-28)
Freshwater prawn Macrobrachium rosenbergii inoculated with 100 μl novel pathogen spiroplasma strain MR-1008 in logarithmic phase (10(8) spiroplasmas ml(-1)) were examined for alkaline phosphatase (AKP) activity, acid phosphatase (ACP) activity, superoxide dismutase (SOD) activity, catalase (CAT) activity, as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service