Skip to Content
Merck
All Photos(1)

Documents

07-1885

Sigma-Aldrich

1,4-Dichlorobenzene

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
1680023
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

SAJ first grade

vapor density

5.07 (vs air)

vapor pressure

1.03 mmHg ( 25 °C)

Assay

≥99.0%

form

sheet

availability

available only in Japan

bp

173 °C (lit.)

mp

52-54 °C (lit.)

density

1.241 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Cl)cc1

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

OCJBOOLMMGQPQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Takahashi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(1), 49-56 (2010-10-12)
Rats and mice (8 animals per species per group) were injected subcutaneously or intraperitoneally with paradichlorobenzene (PDCB) at doses of 0, 100, 200 and 400 mg/kg/day, 4-5 days a week, for 8 weeks (for rats) and either 2 or 6
Gergana Koleva et al.
Journal of the American Chemical Society, 133(47), 19094-19101 (2011-11-09)
The electrophilic sulfonation of several arenes with SO(3) was examined by electronic structure computations at the M06-2X/6-311+G(2d,2p) and SCS-MP2/6-311+G(2d,2p) levels of theory. In contrast to the usual interpretations, the results provide clear evidence that in nonpolar media and in the
Michael C McCarthy et al.
Environmental health perspectives, 117(5), 790-796 (2009-05-30)
Ambient measurements of hazardous air pollutants (air toxics) have been used to validate model-predicted concentrations of air toxics but have not been used to perform risk screening at the national level. We used ambient concentrations of routinely measured air toxics
J A Barter et al.
Regulatory toxicology and pharmacology : RTP, 29(1), 64-79 (1999-03-03)
1,4-Dichlorobenzene (1,4-DCB) was shown to induce the formation of male rat renal tubule tumors and male and female mouse liver tumors when administered in a chronic bioassay. Since the original carcinogenicity findings, an extensive body of mechanistic information has been
Diana E Hun et al.
Environmental health perspectives, 117(12), 1925-1931 (2010-01-06)
Hispanics are the fastest growing minority group in the United States; however, minimal information is available on their cancer risks from exposures to hazardous air pollutants (HAPs) and how these risks compare to risks to non-Hispanic whites. We estimated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service