Skip to Content
Merck
All Photos(1)

Documents

07-1090

Sigma-Aldrich

Diatomaceous earth non-washed

CP

Synonym(s):

Diatomaceous earth, Diatomaceous silica, Kieselguhr

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Highly insoluble, inert material for use as a filtration aid.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C D Martin et al.
Medical and veterinary entomology, 26(3), 323-333 (2012-03-31)
Hen housing (cage or cage-free) did not impact overall abundances of northern fowl mites, Ornithonyssus sylviarum (Canestrini & Fanzago) (Acari: Macronyssidae), or chicken body lice, Menacanthus stramineus (Nitzsch) (Phthiraptera: Menoponidae). Cage-free hens received a dustbox with sand plus diatomaceous earth
Goran G Andrić et al.
Journal of economic entomology, 105(2), 670-678 (2012-05-23)
Insecticidal potential of natural zeolites and diatomaceous earths originating from Serbia against Sitophilus oryzae (L.) and Tribolium castaneum (Herbst) was evaluated. Two natural zeolite formulations (NZ and NZ Modified) were applied to wheat at rates of 0.50, 0.75, and 1.0
James F Meadow et al.
Microbial ecology, 65(1), 171-179 (2012-08-29)
Geothermal influenced soils exert unique physical and chemical limitations on resident microbial communities but have received little attention in microbial ecology research. These environments offer a model system in which to investigate microbial community heterogeneity and a range of soil
Trevor A Lambkin et al.
Journal of economic entomology, 105(3), 979-987 (2012-07-21)
Spinosad, diatomaceous earth, and cyfluthrin were assessed on two broiler farms at Gleneagle and Gatton in southeastern Queensland, Australia in 2004-2005 and 2007-2009, respectively to determine their effectiveness in controlling lesser mealworm, Alphitobius diaperinus (Panzer) (Coleoptera: Tenebrionidae). Insecticide treatments were
Tino Jaschinski et al.
Rapid communications in mass spectrometry : RCM, 27(1), 109-116 (2012-12-15)
Organic matrices are the state-of-the-art ionization mediators in Laser Desorption/Ionization Mass Spectrometry (LDI-MS). Despite improvements in understanding matrix chemistry, interfering matrix-related signals complicate the analysis. Surface-assisted LDI techniques like desorption/ionization on silicon (DIOS) or nanostructure initiator mass spectrometry (NIMS) provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service