Skip to Content
Merck
All Photos(1)

Key Documents

01-6100

Sigma-Aldrich

Antimony standard solution

1000 ppm Sb, suitable for atomic absorption spectrometry

Synonym(s):

Antimony(III) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1000 ppm Sb

suitability

suitable for atomic absorption spectrometry

SMILES string

[Sb]

InChI

1S/Sb

InChI key

WATWJIUSRGPENY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Gurnani et al.
Cytobios, 70(281), 131-136 (1992-01-01)
Clastogenic effects of antimony trichloride, used in small industries, were monitored in laboratory bred white Swiss mice in vivo, following oral administration by gavaging, after 6, 12, 18 and 24 h. Chromosomal aberrations were principally breaks and damaged cells observed
T Gebel et al.
Anticancer research, 18(6A), 4253-4257 (1999-01-19)
Antimony compounds are supposed to resemble to arsenicals in some toxicological features. Comparative investigations with antimony and arsenic were performed to collect data on the genotoxicity of antimony, on which the knowledge is scarce. In comparison to trivalent arsenic, trivalent
S A Hussain et al.
Human & experimental toxicology, 17(3), 140-143 (1998-05-20)
1. The toxic gas hypothesis proposes exposure to stibine (antimony trihydride) generated from microbial contamination of cot mattress materials as a possible cause of unexplained death in infancy (SIDS) as a consequence of cholinesterase inhibition. We have measured the direct
Why not Nichols chemosurgery?
Gary A Dyer
Clinics in dermatology, 24(5), 458-460 (2006-09-13)
The response of different photometers to the color produced by vitamin A and carotene with antimony trichloride.
M J CALDWELL et al.
Transactions of the Kansas Academy of Science. Kansas Academy of Science, 49(2), 197-204 (1946-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service