Skip to Content
Merck
All Photos(1)

Key Documents

PWOPOL-RO

Roche

Pwo DNA Polymerase

Synonym(s):

polymerase, dna, pwo

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106300

biological source

microbial (Pyrococcus woesei)

Quality Level

form

liquid

usage

sufficient for ≤200 reactions (11644955001)
sufficient for 200 reactions
sufficient for ≤40 reactions (11644947001)
sufficient for 40 reactions

specific activity

≥5000 U/mL

mol wt

90 kDa

feature

High Fidelity PCR
dNTPs included: no
hotstart: no

packaging

pkg of 100 U (11644947001)
pkg of 500 U (11644955001 [2 x 250 U])

manufacturer/tradename

Roche

concentration

2.5 units/reaction

technique(s)

PCR: suitable

color

colorless

input

purified DNA

solubility

water: soluble

suitability

suitable for enzyme test

UniProt accession no.

application(s)

genomic analysis
life science and biopharma

foreign activity

Endonucleases with lambda-DNA 30 units, none detected
Nicking act using pBR322-DNA ≤30 units, none detected

storage temp.

−20°C

General description

Pwo DNA Polymerase is a highly processive thermostable 5′-3′ DNA polymerase and possesses a 3′–5′ exonuclease activity, also known as proofreading activity. The enzyme has no detectable 5′–3′ exonuclease activity. Pwo DNA Polymerase exhibits increased thermal stability with a half-life of more than two hours at +100 °C, compared to Taq DNA Polymerase′s half-life of less than 5 minutes at this temperature. Pwo DNA Polymerase-generated PCR products are blunt-ended and can therefore be used directly for blunt-end ligation without any pretreatment of the ends.
The use of Pwo DNA Polymerase during PCR significantly reduces the occurrence of random amplification errors.

Application

  • Due to its proofreading activity, the thermostable Pwo DNA Polymerase has an extremely low error rate, 18-fold lower compared to Taq DNA Polymerase. It is therefore ideal for applications that require the highest possible fidelity in DNA synthesis. It can be applied for High fidelity PCR
  • Cloning of PCR products
  • Characterization of rare mutations
  • PCR

Features and Benefits

  • Excellent accuracy (18-fold more accurate than Taq DNA polymerase)
  • High thermal stability
  • Nearly as processive as Taq DNA polymerase
  • Accepts modified nucleotides

Packaging

1 kit containing 4 components

Quality

Each lot of Pwo DNA polymerase is assayed for:
  • Activity: The enzyme is tested on activated DNA.
  • Function: The enzyme is tested in two PCRs, using λDNA and human genomic DNA as templates.
  • Proofreading ability: Proofreading activity is assayed according to the laq Iq fidelity assay [Frey, B. & Suppmann, B. (1995) Biochemica 2. 8-9].
  • Absence of nucleases: The enzyme is tested on various substrates to ensure the absence of detectable endonucleases, exonucleases, and nicking activity according to the current Quality Control procedures.

Unit Definition

One unit Pwo DNA polymerase is defined as the amount of enzyme that catalyzes the incorporation of 10 nmol total deoxynucleoside triphosphates into acid precipitable DNA within 30 minutes at +70 °C under the conditions described below.

Unit Assay: Incubation buffer for assay on activated DNA

20 mM Tris-HCl, pH 8.8 (20 °C), 50 mM KCl, 2.5 mM MgCl2, 10 mM 2-mercaptoethanol, 0.2 mM of each dATP, dCTP, dGTP, dTTP.


Incubation procedure

12.5 mg activated calf thymus DNA and 0.1 mCi [α-32P]dCTP are incubated with 0.01 to 0.1 U Pwo DNA Polymerase in 50 μl incubation buffer with a paraffin oil overlay at +70 °C for 30 minutes. The amount of incorporated dNTPs is determined by trichloroacetic acid precipitation followed by scintillation counting.

Volume Activity: 5 U/μl

Preparation Note

Pwo DNA polymerase was originally isolated from Pyrococcus woesei, a hyperthermophilic archaebacterium.
Modified nucleotides are substrates
Pwo DNA Polymerase accepts modified nucleotides like digoxigenin-dUTP, biotin-dUTP, or fluorescein-dUTP. Thus, it can add these nucleotides to DNA during PCR. These nonradioactively labeled products can be used as a hybridization probe in many applications.
Magnesium concentration
If you use the magnesium-containing reaction buffer supplied with the enzyme, the final MgCl2 concentration in the PCR will be 2.0mM. For other magnesium concentrations (e.g., for optimizing the reaction to accommodate a particular template), use the magnesium-free reaction buffer and add appropriate amounts of the magnesium stock.

Storage and Stability

Containers which are opened must be carefully resealed and
kept upright to prevent leakage

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

Use of this product is covered by US patent claims and corresponding patent claims outside the US. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patent claims require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.</ br>NOTICE TO PURCHASER: LIMITED LICENSE<br />Use of this product is covered by US Patent No. 6,127,155 and corresponding patent claims outside the US.  The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research.  No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel.  This product is for research use only. Human and veterinary diagnostic uses under Roche patent claims require a separate license from Roche.  All uses other than internal research and human and veterinary diagnostic uses under Roche patent claims require a separate license from Thermo Fisher Scientific. Further information on purchasing licenses from Roche may be obtained by contacting the Licensing Department of Roche Molecular Systems, Inc., 4300 Hacienda Drive, Pleasanton, California 94588, USA or Roche Diagnostics GmbH, Sandhofer Strasse 116, 68305 Mannheim, Germany.  Further information on purchasing licenses from Thermo Fisher Scientific may be obtained by contacting the Licensing Department of  Thermo Fisher Scientific, 5791 Van Allen Way, Carlsbad, California 92008, USA.

Kit Components Only

Product No.
Description

  • Enzyme is supplied in storage and dilution buffer

  • PCR buffer, with 20 mM MgSO4 10x concentrated

  • PCR buffer, without MgSO4 10x concentrated

  • MgSO4 stock solution

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of two rate-limiting steps in the degradation of partially folded immunoglobulin light chains
Mann, et al.
Frontiers in Cell and Developmental Biology, 10 (2022)
General method for site-directed mutagenesis
R Andag and E Schutz
Biotechniques, 30(3), 486-488 (2001)
Insights into the heparan sulphate-dependent externalisation of transglutaminase-2 (TG2) in glucose-stimulated proximal-like tubular epithelial cells
Furini, et al.
Analytical Biochemistry, 603 (2020)
Increased On-Target Rate and Risk of Concatemerization after CRISPR-Enhanced Targeting in ES Cells
Erbs, et al.
Genes, 14 (2023)
Mike McPherson and Simon M?ller
PCR null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service