Skip to Content
Merck
All Photos(1)

Documents

PROTAA-RO

Roche

Protein A Agarose

>98% (HPLC and SDS-PAGE), suspension

Synonym(s):

agarose, protein a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133

Assay

>98% (HPLC and SDS-PAGE)

form

suspension

packaging

pkg of 15 mL (05015979001 [settled resin volume])
pkg of 2 mL (11719408001 [settled resin volume])
pkg of 5 mL (11134515001 [settled resin volume])

manufacturer/tradename

Roche

capacity

>18 mg/mL binding capacity (human IgG (polyclonal))(the IgG is loaded at pH 7.4 and eluted with 200 mM glycine, pH 2.0)

storage temp.

2-8°C

General description

The use of recombinant Protein A in the production of Protein A agarose has the advantage that it is entirely free from Staphylococcal enterotoxins that are known to be mitogenic, and are contaminants in Protein A isolated from S. aureus. The immobilization chemistry provides high IgG-binding capacity and leak-proof stability under a wide range of pH and buffer conditions. The gel can be used over 30 times, depending on the elution condition.

Protein A leakage: <18 ng Protein A/ml (ELISA)
Regeneration: the gel can be used approximately 30 times
Structure: recombinant Protein A (E. coli, Mr = 45,000) is covalently coupled to crosslinked 6% agarose beads: 3 mg Protein A (>98% pure, HPLC, SDS-Page)/1 ml gel
Protein A, immobilized.

Specificity

Protein A is a cell wall protein, isolated from a specific bacterial strain, which has specific binding sites for certain classes of immunoglobulins from different species. Protein A binds to varying degrees IgM, IgA, IgD, and most subclasses of IgG.

Application

Purification of mouse monoclonal antibodies from ascites fluid or cell culture supernatants. Protein A or Protein G Agarose affinity matrix can be used to purify antibodies from crude cell extracts and for the immunoprecipitation or co-immunoprecipitation of proteins. It has also been used for photoactivatable-ribonucleoside-enhanced cross-linking immunoprecipitation (PAR-CLIP) analysis.

Components

Preswollen gel, ready-to-use, 2 ml or 5 ml gel (settled gel volume) in buffer that contains 20% ethanol as a preservative, nonsterile.
Protein A content: 3 mg/ml pre-swollen gel

Quality

Purity: >98% (HPLC, SDS-PAGE); entirely free from staphylococcal enterotoxins.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiuqing He et al.
Cells, 12(12) (2023-06-28)
The axoneme and accessory structures of flagella are critical for sperm motility and male fertilization. Sperm production needs precise and highly ordered gene expression to initiate and sustain the many cellular processes that result in mature spermatozoa. Here, we identified
Vladimir V Rogov et al.
EMBO reports, 18(8), 1382-1396 (2017-06-29)
Through the canonical LC3 interaction motif (LIR), [W/F/Y]-X1-X2-[I/L/V], protein complexes are recruited to autophagosomes to perform their functions as either autophagy adaptors or receptors. How these adaptors/receptors selectively interact with either LC3 or GABARAP families remains unclear. Herein, we determine
David Llobet-Navas et al.
Molecular and cellular biology, 34(23), 4216-4231 (2014-10-01)
Recently, we demonstrated that the microRNA 424(322)/503 [miR-424(322)/503] cluster is transcriptionally controlled by transforming growth factor β (TGF-β) in the mammary epithelium. Induction of this microRNA cluster impacts mammary epithelium fate by regulating apoptosis and insulin-like growth factor 1 (IGF1)
R Travis Taylor et al.
Methods (San Diego, Calif.), 55(2), 166-171 (2011-08-23)
Ubiquitin (Ub) conjugation to a substrate protein is a widely used cellular mechanism for control of protein stability and function, modulation of signal transduction pathways and antiviral responses. Identification and characterization of ubiquitinated viral proteins is an important step in
Florian Dunker et al.
Bio-protocol, 11(3), e3911-e3911 (2021-03-19)
Over the last decade, it has been noticed that microbial pathogens and pests deliver small RNA (sRNA) effectors into their host plants to manipulate plant physiology and immunity for infection, known as cross kingdom RNA interference. In this process, fungal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service