Skip to Content
Merck
All Photos(1)

Key Documents

EAGLETMMRO

Roche

EagleTaq Universal Master Mix (ROX)

sufficient for 100 reactions, sufficient for 500 reactions, sufficient for 5000 reactions, suitable for qPCR, suitable for RT-qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for 100 reactions
sufficient for 500 reactions
sufficient for 5000 reactions

Quality Level

feature

dNTPs included
hotstart

manufacturer/tradename

Roche

packaging

pkg of 100 x 20 μL reactions (07260288190)
pkg of 500 x 20 μL reactions (07260296190)
pkg of 5000 x 20 μL reactions (07249926190)

technique(s)

RT-qPCR: suitable
qPCR: suitable

input

purified DNA

detection method

probe-based

General description

2× concentrated, ready-to-use hot start master mix for qPCR and qRT-PCR using the hydrolysis probe detection format on real-time PCR instruments.

The EagleTaq Universal Master Mix (ROX) is a ready-to-use, 2× concentrated PCR master mix that contains all the reagents (except primers, probes, and template) needed for performing quantitative, real-time PCR hydrolysis probe reactions. It contains a special ROX reference dye which makes it suitable for all real-time instruments on which a ROX reference dye is needed for quantitative analysis.
The EagleTaq Universal Master Mix (ROX) contains dUTP so that it may be used with Uracil-DNA Glycosylase (UNG) to prevent false positives arising from carryover contamination.
The hot start properties allow reaction setup at ambient temperature.
With this robust reagent, any PCR protocol optimization is minimized
The EagleTaq Universal Master Mix (ROX) is a ready-to-use, 2× concentrated polymerase chain reaction (PCR) master mix that contains all the reagents (except primers, probes, and template) needed for performing quantitative, real-time PCR (qPCR) and quantitative reverse transcription PCR (qRT-PCR) hydrolysis probe reactions.

Application

EagleTaq Universal Master Mix (ROX) has been used in quantitative, real-time polymerase chain reaction (qPCR ) and quantitative reverse transcription PCR (qRT-PCR).

Features and Benefits

  • The EagleTaq Universal Master Mix (ROX) contains a special ROX reference dye which makes it suitable for all real-time instruments on which a ROX reference dye is needed for quantitative analysis.
  • It contains dUTP so that it may be used with Uracil-DNA Glycosylase (UNG) to prevent false positives arising from carryover contamination.
  • The hot start properties allow reaction setup at ambient temperature.
  • With this robust reagent, any PCR protocol optimization is minimized.

Components

Contains EagleTaq DNA Polymerase and a reaction buffer with dATP, dCTP, dGTP, dUTP, MgCl2, and ROX.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroto Suwa et al.
Biochemistry and biophysics reports, 8, 62-67 (2016-08-12)
Retinoic acid (RA) is a metabolite of vitamin A and has important roles in development, differentiation, and reproduction. Activin has been shown to regulate the RA pathway and affect granulosa cell (GC) proliferation, suggesting that RA is important for early
Ai Nakaoka et al.
Oncology reports, 45(4) (2021-03-03)
The precise mechanism of intercellular communication between cancer cells following radiation exposure is unclear. Exosomes are membrane‑enclosed small vesicles comprising lipid bilayers and are mediators of intercellular communication that transport a variety of intracellular components, including microRNAs (miRNAs or miRs).
James Meixiong et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(21), 10525-10530 (2019-05-10)
Patients suffering from cholestasis, the slowing or stoppage of bile flow, commonly report experiencing an intense, chronic itch. Numerous pruritogens are up-regulated in cholestatic patient sera, including bile acids (BAs). Acute injection of BAs results in itch in both mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service