Skip to Content
Merck
All Photos(1)

Key Documents

11218590910

Roche

DNA Molecular Weight Marker II, DIG-labeled

greener alternative

solution, pkg of 500 μL (10 μg/ml)

Synonym(s):

DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

Quality Level

packaging

pkg of 500 μL (10 μg/ml)

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10 μg/mL

color

colorless

solubility

water: miscible

greener alternative category

storage temp.

−20°C

General description

DNA Molecular Weight Marker II, DIG-labeled is a solution in 10 mM Tris-HCl, 1 mM EDTA, pH 8.0. This fragment mixture is prepared by cleavage of λDNA with Hind III. The size of the fragment ranges from 0.12 to 23.1 kbp. A photodigoxigenin has been introduced at approx. every 200th to 300th base in the DNA fragments.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Application

DNA Molecular Weight Marker II, DIG-labeled has been used as a size standard in Southern blot analysis for nucleic acid labeling and detection.

Features and Benefits

Contents
Ready-to-use solutions in 10mM Tris-HCl, 1mM EDTA, pH 8.0.

Sequence

The mixture contains 8 fragments with the following base pair lengths: 125, 564, 2027, 2322, 4361, 6557, 9416, and 23130 bp.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The APETALA1 and FRUITFUL homologs in Camellia japonica and their roles in double flower domestication
Yingkun Sun
Molecular breeding : new strategies in plant improvement, 33(4), 821-834 (2014)
Yingkun Sun et al.
BMC plant biology, 14, 288-288 (2014-10-26)
Double flower domestication is of great value in ornamental plants and presents an excellent system to study the mechanism of morphological alterations by human selection. The classic ABC model provides a genetic framework underlying the control of floral organ identity
Haipeng Xue et al.
Methods in molecular biology (Clifton, N.J.), 1307, 173-190 (2014-03-13)
Genetic modification is an indispensable tool to study gene function in normal development and disease. The recent breakthrough of creating human induced pluripotent stem cells (iPSCs) by defined factors (Takahashi et al., Cell 131:861-872, 2007) provides a renewable source of
Yongquan Luo et al.
Current protocols in stem cell biology, 29, 5A-5A (2014-05-20)
Generation of a fluorescent GFP reporter line in human induced pluripotent stem cells (hiPSCs) provides enormous potentials in both basic stem cell research and regenerative medicine. A protocol for efficiently generating such an engineered reporter line by gene targeting is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service