Skip to Content
Merck
All Photos(2)

Key Documents

03353591910

Roche

DIG Gel Shift Kit, 2nd generation

greener alternative

storage temp.:-20°C (-15°C to -25°C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.55

description

New formulation containing Terminal Transferase, recombinant.

Quality Level

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

General description

DIG Gel Shift Kit, 2nd generation contains reagents for nonradioactive 3′-end labeling of oligonucleotides, to be used in a "gel mobility shift" assay. It also contains offers, electrophoresis reagents and an enzyme-labeled antibody to facilitate the detection of DNA-protein complexes.
DIG Gel Shift Kit employs recombinant terminal transferase and digoxigenin (DIG)-11- dideoxyuridine triphosphate (ddUTP) makes the labeling reaction very flexible. It can be used to label the 3′ ends of any oligonucleotide (whether it has a 5′- overhanging end, a 3′-overhanging end, or blunt ends). Both single- and double-stranded DNA can be labeled.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Application

DIG Gel Shift Kit, 2nd generation has been used for nonradioactive detection of sequence-specific DNA binding proteins. The DIG Gel Shift Kit is used to prepare nonradioactive, 3′-end labeled oligonucleotide probes to detect DNA-protein complexes in a "gel mobility shift assay".

Features and Benefits

The electrophoresis assay works best with shorter (30 to 100bp) DNA fragments, since these are less likely to have sequences that are outside the specific binding site, but can interact nonspecifically with target proteins.
  • Convenient, since the technique does not require special equipment or technology.
  • Reproducible, since DIG-labeled probes are stable indefinitely.
  • Safe, because the assay is nonradioactive.
  • Reliable, because the kit provides DIG-labeled control oligonucleotides to establish that the assay is working.
  • Function-tested with the controls provided in the kit (See "Quality").
  • Sensitive, since the kit can detect as little as 20fmol of the control oligonucleotide (after it is labeled according to the kit protocol).

Packaging

1 kit containing 16 components. 1 kit for up to 20 Oligonucleotide 3-end labeling reactions with DIG-11-ddUTP, 200 binding reactions, chemiluminescent detection reaction for up to 20 blots, DNA binding protein and oligonucleotide for up to 20 control reactions

Physical form

Chlorophenol red-β-D-galactopyranoside, monosodium salt

Preparation Note

The study of DNA-protein interactions has been greatly facilitated in recent years by the rapid and simple "gel retardation" or "gel mobility shift" assay. Because free DNA and DNA-protein complexes migrate differently during gel electrophoresis, they can be separated and detected on native (nondenaturing) polyacrylamide or agarose gels.

Sample
Amount: 3.85pmol or 100ng
Type: Oligonucleotides with 5′-overhanging ends, 3′-overhanging ends, or blunt ends; single- or double-stranded DNA fragments (30 - 200bp)
Note: Ideally, fragments to be labeled should be between 30 and 100bp.
Time required
Oligonucleotide annealing and labeling: 10minutes
Formation of oligonucleotide-protein complexes: 25minutes
Electrophoresis: 1 hour to overnight, depending on electrophoresis system
Blotting: 1 - 2 hours
Immunological detection: 2hours
Exposure to X-ray film or imager: 15 - 40minutes

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Labeling Buffer 5x concentrated

  • CoCl<SUB>2</SUB> Solution 25 mM

  • DIG-ddUTP Solution, 1 mM Digoxigenin-11-ddUTP

  • Recombinant Terminal Transferase 400 U/μl

  • Binding Buffer 5x concentrated

  • Control Oligonucletide ds 39mer, unlabeled 0.1 μg/μl, 3.85 pmol/μl

  • DIG-labeled Control Oligonucleotide ds 39mer 0.4 ng/μl, 15.54 fmol/μl

  • Control Factor Oct2A 25-75 ng/μl

  • Poly [d(I-C)] 0.1 μg/μl

  • Poly [d(A-T)] 0.1 μg/μl

  • Poly L-lysine 0.1 μg/μl

  • Loading Buffer without bromophenol blue

  • Loading Buffer with bromophenol blue

  • Anti-Digoxigenin-AP 750 U/ml

  • CSPD 10 mg/ml

  • Blocking Reagent

See All (16)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B Inhalation - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunhua Ding et al.
Molecular plant, 12(9), 1259-1277 (2019-05-28)
RNA polymerase transcriptional pausing represents a major checkpoint in transcription in bacteria and metazoans, but it is unknown whether this phenomenon occurs in plant organelles. Here, we report that transcriptional pausing occurs in chloroplasts. We found that mTERF5 specifically and
Ravindra B Chalamalasetty et al.
Development (Cambridge, England), 141(22), 4285-4297 (2014-11-06)
Neuromesodermal (NM) stem cells generate neural and paraxial presomitic mesoderm (PSM) cells, which are the respective progenitors of the spinal cord and musculoskeleton of the trunk and tail. The Wnt-regulated basic helix-loop-helix (bHLH) transcription factor mesogenin 1 (Msgn1) has been
Erchao Duan et al.
The Plant cell, 31(5), 1026-1042 (2019-03-28)
Tillering and panicle branching are important determinants of plant architecture and yield potential in rice (Oryza sativa). IDEAL PLANT ARCHITECTURE1 (IPA1) encodesSQUAMOSA PROMOTER BINDING PROTEIN-LIKE14, which acts as a key transcription factor regulating tiller outgrowth and panicle branching by directly
Os PEX 5 regulates rice spikelet development through modulating jasmonic acid biosynthesis
<BIG>You X, et al.</BIG>
The New phytologist, 224, 712-724 (2019)
Ting Lei et al.
Virulence, 6(1), 75-84 (2014-12-18)
The staphylococcal YeaZ is highly conserved in prokaryotic cells and critical for growth of many bacterial pathogens. However, the essentiality for Staphylococcus aureus growth and the biological function of YeaZ behind its essentiality remain undefined. In this study, we created

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service