ZUFPREUN0
UF Pretreatment Unit 3/4
Removes contaminants from tap water with fouling index above system specifications
All Photos(1)
About This Item
AC/DC input
6 V
packaging
pkg of 1 unit
parameter
22 kg operating weight (48.5 lb)
unit H × W × D
129 cm (50.8 in.) × 30 cm (11.8 in.) × 25 cm (9.8 in.)
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
This ultrafiltration (UF) pretreatment unit removes suspended particulates, microorganisms, and colloids from a tap water and is used when tap water has a high Fouling Index (7 > SDI > 12).
Application
Pretreats tap water with high fouling index before being fed to Milli-Q® CLX/HX/HX-SD/HR 7000 systems.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany