Skip to Content
Merck
All Photos(1)

Key Documents

ZSQ240CTT0

Milli-Q

Milli-Q® SQ Cleaning Tool

Sanitization of the Milli-Q® SQ Switch Tank and Milli-Q® SQ/SR production station

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
NC.75

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® SQ 2Series

General description

To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The Milli-Q® SQ Cleaning Tool is a specially dedicated tool for the sanitization of the Milli-Q® SQ/SR production station and its Milli-Q® SQ Switch Tank. It uses chlorine tablets, such ROProtect C or EfferSan (not included) for the sanitization process. There is a specific procedure to be followed for the usage of this tool.

Components

The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice. This catalog number does not include ROProtect C and EfferSan Effervescent Chlorine tablets.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service