ZAFS52120
Milli-Q® CLX Water Purification System
Delivers up to 2400 L/day of clinical laboratory reagent water (CLRW)
Synonym(s):
Milli-Q® CLX 7120 Water Purification System 230V 50/60 Hz
About This Item
Recommended Products
AC/DC input
100 V / 230 V (For region specific voltage information, see footnote below)
Quality Level
product line
Milli-Q® CLX
packaging
pkg of 1 unit
parameter
120 L/hr make-up flow rate
2400 L/day max. usage
303 kg operating weight (668 lb)
4 L/min distribution flow rate
resistivity
>15 MΩ-cm, at 25°C
H × W × depth
125.5 cm (48.8 in.) × 54.3 cm (21.4 in.) × 94.7 cm (37.3 in.)
total organic carbon (TOC) residue
<30 ppb (Product water)
input
potable tap water
output
clinical laboratory reagent water (CLRW) (complies with the CLSI guideline)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- 24/7 real-time monitoring and secure remote-control access.
- Automatic e-record archiving for up to two years facilitates laboratory accreditation and reaccreditation processes.
- Patented Elix® electrodeionization (EDI) module provides consistently high-quality water with no need for resin cylinders. This reduces maintenance, running costs and analyzer downtime.
- Unique E.R.A.® technology automatically optimizes water recovery based on feed water quality to decrease water usage costs.
- Three built-in bactericidal UV lamps reduce system and analyzer sanitization needs to maximize lab uptime.
- A large, user-friendly color touchscreen shows all important system information at a glance and allows for intuitive operation.
- To ensure you always have an uninterrupted supply of pure water, these robust systems are equipped with an emergency bypass feature that is easy to set up.
- The systems are compact and can be installed wherever it is convenient in the laboratory. They are also customizable, allowing you to adapt them to your specific requirements.
- Available with MyMilli-Q™ Remote Care monitoring and service to support maximum uptime.
Components
- The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
- This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
- Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!
Other Notes
Organism Retention: Microorganisms
Mode of Action: Filtering and UV
Application: Clinical laboratories
Intended Use: Water purification
Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes and exposed to bactericidal UV lamp. Please refer to system equipment user guide section "System" (provided on USB key in the North American configuration version).
Storage Statement: Store in dry location
Disposal Statement: Dispose of in accordance with applicable federal, state, and local regulations.
Footnote
- 230 V systems are required for Europe, Africa and Middle East regions, and certain LATAM and APAC countries
- 120 V systems are required for North America, Taiwan, and certain LATAM countries
- 110 V systems are required for Japan
Legal Information
Disclaimer

Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service